Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42744257-42744767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

SNP Detail

SNP ID Chromosome Location
rs902977121 Chr.15: 42744453-42744453 on Build GRCh38
rs560041206 Chr.15: 42744690-42744690 on Build GRCh38
rs928707597 Chr.15: 42744299-42744299 on Build GRCh38
rs148272721 Chr.15: 42744484-42744484 on Build GRCh38
rs1029043954 Chr.15: 42744337-42744338 on Build GRCh38
rs1036601048 Chr.15: 42744322-42744322 on Build GRCh38
rs900106446 Chr.15: 42744585-42744585 on Build GRCh38
rs1035802639 Chr.15: 42744691-42744691 on Build GRCh38
rs561020553 Chr.15: 42744524-42744524 on Build GRCh38
rs574886759 Chr.15: 42744449-42744449 on Build GRCh38
rs754871195 Chr.15: 42744607-42744607 on Build GRCh38
rs746843462 Chr.15: 42744575-42744575 on Build GRCh38
rs916538375 Chr.15: 42744628-42744628 on Build GRCh38
rs368522546 Chr.15: 42744720-42744720 on Build GRCh38
rs781080005 Chr.15: 42744642-42744642 on Build GRCh38
rs539070978 Chr.15: 42744407-42744407 on Build GRCh38
rs1008579524 Chr.15: 42744395-42744395 on Build GRCh38
rs1051556789 Chr.15: 42744506-42744506 on Build GRCh38
rs940830656 Chr.15: 42744651-42744651 on Build GRCh38
rs79592696 Chr.15: 42744633-42744633 on Build GRCh38
rs779658288 Chr.15: 42744557-42744557 on Build GRCh38
rs541966678 Chr.15: 42744681-42744681 on Build GRCh38
rs888802755 Chr.15: 42744386-42744386 on Build GRCh38
rs982171510 Chr.15: 42744641-42744641 on Build GRCh38
rs76933141 Chr.15: 42744347-42744347 on Build GRCh38
rs995736107 Chr.15: 42744587-42744587 on Build GRCh38
rs562554712 Chr.15: 42744300-42744300 on Build GRCh38
rs947260312 Chr.15: 42744335-42744335 on Build GRCh38
rs771317015 Chr.15: 42744433-42744433 on Build GRCh38
rs529718030 Chr.15: 42744532-42744532 on Build GRCh38
rs1017200168 Chr.15: 42744574-42744574 on Build GRCh38
rs890189498 Chr.15: 42744545-42744545 on Build GRCh38
rs977259448 Chr.15: 42744490-42744490 on Build GRCh38
rs1032889700 Chr.15: 42744534-42744534 on Build GRCh38
rs1043290289 Chr.15: 42744418-42744418 on Build GRCh38
rs1055787292 Chr.15: 42744302-42744302 on Build GRCh38
rs571544379 Chr.15: 42744332-42744332 on Build GRCh38

View Full Product Details