Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42753037-42753476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3886995 c.2197C>T p.Q733* Chr.15: 42753049-42753049 on Build GRCh38
4054755 c.2176A>G p.S726G Chr.15: 42753070-42753070 on Build GRCh38
3501330 c.2120C>T p.P707L Chr.15: 42753126-42753126 on Build GRCh38
3501331 c.2119C>T p.P707S Chr.15: 42753127-42753127 on Build GRCh38
297213 c.2023G>T p.A675S Chr.15: 42753223-42753223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758080551 Chr.15: 42753275-42753275 on Build GRCh38
rs367778872 Chr.15: 42753089-42753089 on Build GRCh38
rs761467140 Chr.15: 42753258-42753258 on Build GRCh38
rs749884465 Chr.15: 42753271-42753271 on Build GRCh38
rs374412910 Chr.15: 42753080-42753080 on Build GRCh38
rs781635869 Chr.15: 42753184-42753184 on Build GRCh38
rs377299672 Chr.15: 42753157-42753157 on Build GRCh38
rs76000738 Chr.15: 42753113-42753113 on Build GRCh38
rs965735530 Chr.15: 42753321-42753321 on Build GRCh38
rs751987449 Chr.15: 42753298-42753298 on Build GRCh38
rs187389348 Chr.15: 42753104-42753104 on Build GRCh38
rs2899061 Chr.15: 42753382-42753382 on Build GRCh38
rs773148128 Chr.15: 42753408-42753408 on Build GRCh38
rs759376111 Chr.15: 42753071-42753071 on Build GRCh38
rs778756599 Chr.15: 42753115-42753115 on Build GRCh38
rs372443449 Chr.15: 42753186-42753186 on Build GRCh38
rs369896751 Chr.15: 42753175-42753175 on Build GRCh38
rs769842511 Chr.15: 42753203-42753203 on Build GRCh38
rs1008775283 Chr.15: 42753412-42753412 on Build GRCh38
rs1044486295 Chr.15: 42753262-42753262 on Build GRCh38
rs769711047 Chr.15: 42753345-42753345 on Build GRCh38
rs752644926 Chr.15: 42753085-42753085 on Build GRCh38
rs756796072 Chr.15: 42753114-42753114 on Build GRCh38
rs750086721 Chr.15: 42753151-42753151 on Build GRCh38
rs747277579 Chr.15: 42753162-42753162 on Build GRCh38
rs936020618 Chr.15: 42753309-42753309 on Build GRCh38
rs758756132 Chr.15: 42753154-42753154 on Build GRCh38
rs1031345907 Chr.15: 42753379-42753379 on Build GRCh38
rs779526746 Chr.15: 42753277-42753277 on Build GRCh38
rs951717096 Chr.15: 42753125-42753125 on Build GRCh38
rs577127450 Chr.15: 42753216-42753216 on Build GRCh38
rs917466002 Chr.15: 42753213-42753213 on Build GRCh38
rs775375916 Chr.15: 42753218-42753218 on Build GRCh38
rs753800281 Chr.15: 42753231-42753231 on Build GRCh38
rs375442471 Chr.15: 42753221-42753221 on Build GRCh38
rs763327774 Chr.15: 42753422-42753424 on Build GRCh38
rs926018763 Chr.15: 42753305-42753305 on Build GRCh38
rs1040691800 Chr.15: 42753446-42753446 on Build GRCh38
rs775554436 Chr.15: 42753265-42753265 on Build GRCh38
rs914391169 Chr.15: 42753437-42753437 on Build GRCh38
rs574962615 Chr.15: 42753326-42753326 on Build GRCh38
rs955748255 Chr.15: 42753417-42753417 on Build GRCh38
rs764164213 Chr.15: 42753223-42753223 on Build GRCh38
rs201524659 Chr.15: 42753075-42753075 on Build GRCh38
rs763235763 Chr.15: 42753207-42753207 on Build GRCh38

View Full Product Details