Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45546472-45546993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
UniGene
Hs.466945
Gene Chromosome Location
Chr.19: 45527767-45584864 on Build GRCh38
Transcripts
NM_001017989.2 NM_025136.3 XM_006723403.3

SNP Detail

SNP ID Chromosome Location
rs184867350 Chr.19: 45546546-45546546 on Build GRCh38
rs936878650 Chr.19: 45546545-45546545 on Build GRCh38
rs978118927 Chr.19: 45546748-45546748 on Build GRCh38
rs966291996 Chr.19: 45546583-45546583 on Build GRCh38
rs1026526376 Chr.19: 45546929-45546929 on Build GRCh38
rs948480364 Chr.19: 45546608-45546608 on Build GRCh38
rs928413229 Chr.19: 45546867-45546867 on Build GRCh38
rs375848972 Chr.19: 45546739-45546739 on Build GRCh38
rs924222763 Chr.19: 45546526-45546526 on Build GRCh38
rs866759120 Chr.19: 45546615-45546615 on Build GRCh38
rs773109015 Chr.19: 45546944-45546944 on Build GRCh38
rs188922477 Chr.19: 45546962-45546962 on Build GRCh38
rs535393059 Chr.19: 45546811-45546811 on Build GRCh38
rs112948303 Chr.19: 45546499-45546499 on Build GRCh38
rs539207846 Chr.19: 45546766-45546766 on Build GRCh38
rs886054504 Chr.19: 45546741-45546741 on Build GRCh38
rs1017726666 Chr.19: 45546577-45546577 on Build GRCh38
rs528812143 Chr.19: 45546612-45546612 on Build GRCh38
rs188413392 Chr.19: 45546706-45546706 on Build GRCh38
rs548194990 Chr.19: 45546688-45546688 on Build GRCh38
rs546383446 Chr.19: 45546522-45546522 on Build GRCh38
rs10422253 Chr.19: 45546724-45546724 on Build GRCh38
rs989715820 Chr.19: 45546817-45546817 on Build GRCh38
rs1019694985 Chr.19: 45546741-45546741 on Build GRCh38
rs570533596 Chr.19: 45546764-45546764 on Build GRCh38
rs896785880 Chr.19: 45546920-45546920 on Build GRCh38
rs984769166 Chr.19: 45546622-45546622 on Build GRCh38
rs560024517 Chr.19: 45546958-45546958 on Build GRCh38
rs552769108 Chr.19: 45546785-45546785 on Build GRCh38
rs369362617 Chr.19: 45546558-45546558 on Build GRCh38
rs1015642963 Chr.19: 45546618-45546618 on Build GRCh38
rs566392487 Chr.19: 45546792-45546792 on Build GRCh38
rs895286170 Chr.19: 45546598-45546598 on Build GRCh38
rs897073785 Chr.19: 45546664-45546665 on Build GRCh38
rs534603782 Chr.19: 45546840-45546840 on Build GRCh38
rs1016246843 Chr.19: 45546949-45546949 on Build GRCh38
rs142898530 Chr.19: 45546747-45546747 on Build GRCh38
rs751011663 Chr.19: 45546585-45546585 on Build GRCh38
rs530660050 Chr.19: 45546712-45546712 on Build GRCh38
rs184177890 Chr.19: 45546960-45546960 on Build GRCh38
rs944035967 Chr.19: 45546500-45546500 on Build GRCh38
rs980622657 Chr.19: 45546718-45546718 on Build GRCh38
rs1008173662 Chr.19: 45546500-45546500 on Build GRCh38
rs995414147 Chr.19: 45546936-45546936 on Build GRCh38
rs111523448 Chr.19: 45546530-45546530 on Build GRCh38
rs369198028 Chr.19: 45546899-45546899 on Build GRCh38
rs181576269 Chr.19: 45546826-45546826 on Build GRCh38
rs945042481 Chr.19: 45546801-45546801 on Build GRCh38
rs542594258 Chr.19: 45546961-45546961 on Build GRCh38
rs950963157 Chr.19: 45546943-45546943 on Build GRCh38
rs147443761 Chr.19: 45546857-45546857 on Build GRCh38
rs1006673214 Chr.19: 45546951-45546951 on Build GRCh38
rs1039208478 Chr.19: 45546855-45546855 on Build GRCh38
rs909296769 Chr.19: 45546664-45546664 on Build GRCh38
rs995360091 Chr.19: 45546687-45546687 on Build GRCh38
rs557129054 Chr.19: 45546896-45546896 on Build GRCh38
rs546386601 Chr.19: 45546948-45546948 on Build GRCh38
rs976294041 Chr.19: 45546588-45546588 on Build GRCh38
rs985535527 Chr.19: 45546729-45546729 on Build GRCh38
rs938392779 Chr.19: 45546883-45546883 on Build GRCh38
rs139757997 Chr.19: 45546888-45546888 on Build GRCh38
rs950795783 Chr.19: 45546620-45546620 on Build GRCh38
rs557041940 Chr.19: 45546496-45546496 on Build GRCh38
rs768694312 Chr.19: 45546816-45546816 on Build GRCh38

View Full Product Details