Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45548114-45548547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
UniGene
Hs.466945
Gene Chromosome Location
Chr.19: 45527767-45584864 on Build GRCh38
Transcripts
NM_001017989.2 NM_025136.3 XM_006723403.3

SNP Detail

SNP ID Chromosome Location
rs558389168 Chr.19: 45548333-45548333 on Build GRCh38
rs902049080 Chr.19: 45548494-45548494 on Build GRCh38
rs919354931 Chr.19: 45548385-45548385 on Build GRCh38
rs371869405 Chr.19: 45548397-45548397 on Build GRCh38
rs747076255 Chr.19: 45548312-45548312 on Build GRCh38
rs538145979 Chr.19: 45548252-45548252 on Build GRCh38
rs373641000 Chr.19: 45548463-45548463 on Build GRCh38
rs1024030760 Chr.19: 45548219-45548219 on Build GRCh38
rs930531316 Chr.19: 45548404-45548404 on Build GRCh38
rs190072684 Chr.19: 45548238-45548238 on Build GRCh38
rs549141863 Chr.19: 45548157-45548157 on Build GRCh38
rs1021931461 Chr.19: 45548247-45548247 on Build GRCh38
rs566934008 Chr.19: 45548208-45548208 on Build GRCh38
rs929352492 Chr.19: 45548399-45548399 on Build GRCh38
rs987308299 Chr.19: 45548262-45548262 on Build GRCh38
rs967970192 Chr.19: 45548230-45548230 on Build GRCh38
rs35111539 Chr.19: 45548360-45548361 on Build GRCh38
rs535986790 Chr.19: 45548237-45548237 on Build GRCh38
rs73568970 Chr.19: 45548149-45548149 on Build GRCh38
rs899117691 Chr.19: 45548396-45548396 on Build GRCh38
rs977581211 Chr.19: 45548233-45548233 on Build GRCh38
rs367895706 Chr.19: 45548206-45548206 on Build GRCh38
rs777979909 Chr.19: 45548266-45548266 on Build GRCh38
rs553386926 Chr.19: 45548468-45548468 on Build GRCh38
rs1009247426 Chr.19: 45548243-45548243 on Build GRCh38
rs886054512 Chr.19: 45548256-45548256 on Build GRCh38
rs539808989 Chr.19: 45548424-45548424 on Build GRCh38
rs1014002317 Chr.19: 45548217-45548217 on Build GRCh38
rs573380409 Chr.19: 45548469-45548469 on Build GRCh38
rs770080585 Chr.19: 45548286-45548286 on Build GRCh38
rs1039269566 Chr.19: 45548487-45548487 on Build GRCh38
rs1034014555 Chr.19: 45548188-45548188 on Build GRCh38
rs908586214 Chr.19: 45548267-45548267 on Build GRCh38
rs992685413 Chr.19: 45548367-45548367 on Build GRCh38
rs960565366 Chr.19: 45548339-45548339 on Build GRCh38
rs35934373 Chr.19: 45548503-45548504 on Build GRCh38
rs796325284 Chr.19: 45548402-45548402 on Build GRCh38
rs1036190955 Chr.19: 45548335-45548335 on Build GRCh38
rs577920303 Chr.19: 45548406-45548406 on Build GRCh38
rs780964248 Chr.19: 45548501-45548501 on Build GRCh38
rs1053663944 Chr.19: 45548510-45548510 on Build GRCh38
rs1033952416 Chr.19: 45548293-45548293 on Build GRCh38
rs1055500477 Chr.19: 45548458-45548458 on Build GRCh38
rs975407912 Chr.19: 45548248-45548248 on Build GRCh38
rs867609793 Chr.19: 45548290-45548290 on Build GRCh38
rs547036498 Chr.19: 45548158-45548158 on Build GRCh38
rs921250655 Chr.19: 45548256-45548256 on Build GRCh38
rs933963129 Chr.19: 45548259-45548259 on Build GRCh38

View Full Product Details