Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42775176-42775741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
267656 c.1953G>A p.T651T Chr.15: 42775180-42775180 on Build GRCh38
3886996 c.1559C>T p.S520F Chr.15: 42775574-42775574 on Build GRCh38
7395541 c.1555G>C p.A519P Chr.15: 42775578-42775578 on Build GRCh38
3361395 c.1903G>T p.D635Y Chr.15: 42775230-42775230 on Build GRCh38
3370170 c.1522G>A p.D508N Chr.15: 42775611-42775611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369686818 Chr.15: 42775515-42775515 on Build GRCh38
rs994405586 Chr.15: 42775336-42775336 on Build GRCh38
rs780631079 Chr.15: 42775455-42775455 on Build GRCh38
rs751818995 Chr.15: 42775535-42775535 on Build GRCh38
rs868814760 Chr.15: 42775562-42775562 on Build GRCh38
rs761082723 Chr.15: 42775487-42775487 on Build GRCh38
rs867840054 Chr.15: 42775686-42775686 on Build GRCh38
rs773259776 Chr.15: 42775414-42775414 on Build GRCh38
rs756004916 Chr.15: 42775538-42775538 on Build GRCh38
rs762573258 Chr.15: 42775285-42775285 on Build GRCh38
rs757048974 Chr.15: 42775685-42775685 on Build GRCh38
rs746073661 Chr.15: 42775247-42775247 on Build GRCh38
rs771555123 Chr.15: 42775588-42775588 on Build GRCh38
rs184167929 Chr.15: 42775311-42775311 on Build GRCh38
rs967280552 Chr.15: 42775527-42775527 on Build GRCh38
rs751510158 Chr.15: 42775313-42775313 on Build GRCh38
rs751622999 Chr.15: 42775637-42775637 on Build GRCh38
rs770799522 Chr.15: 42775553-42775553 on Build GRCh38
rs753609835 Chr.15: 42775660-42775660 on Build GRCh38
rs769690874 Chr.15: 42775403-42775403 on Build GRCh38
rs547196103 Chr.15: 42775289-42775289 on Build GRCh38
rs749057161 Chr.15: 42775542-42775542 on Build GRCh38
rs779249410 Chr.15: 42775238-42775238 on Build GRCh38
rs922063750 Chr.15: 42775665-42775665 on Build GRCh38
rs376556189 Chr.15: 42775488-42775488 on Build GRCh38
rs754164056 Chr.15: 42775502-42775502 on Build GRCh38
rs376543555 Chr.15: 42775204-42775204 on Build GRCh38
rs755173026 Chr.15: 42775650-42775650 on Build GRCh38
rs764707596 Chr.15: 42775321-42775321 on Build GRCh38
rs776578713 Chr.15: 42775258-42775258 on Build GRCh38
rs772993319 Chr.15: 42775278-42775278 on Build GRCh38
rs754283212 Chr.15: 42775356-42775356 on Build GRCh38
rs747997967 Chr.15: 42775396-42775396 on Build GRCh38
rs779918165 Chr.15: 42775228-42775228 on Build GRCh38
rs577577749 Chr.15: 42775635-42775635 on Build GRCh38
rs367953866 Chr.15: 42775394-42775394 on Build GRCh38
rs762195077 Chr.15: 42775632-42775632 on Build GRCh38
rs897463629 Chr.15: 42775695-42775695 on Build GRCh38
rs963464682 Chr.15: 42775647-42775647 on Build GRCh38
rs762238899 Chr.15: 42775504-42775504 on Build GRCh38
rs200773248 Chr.15: 42775197-42775197 on Build GRCh38
rs745797698 Chr.15: 42775698-42775698 on Build GRCh38
rs555093202 Chr.15: 42775480-42775480 on Build GRCh38
rs186720367 Chr.15: 42775362-42775362 on Build GRCh38
rs760320746 Chr.15: 42775606-42775606 on Build GRCh38
rs200585314 Chr.15: 42775613-42775613 on Build GRCh38
rs772352231 Chr.15: 42775253-42775253 on Build GRCh38
rs780473530 Chr.15: 42775532-42775532 on Build GRCh38
rs377402244 Chr.15: 42775651-42775651 on Build GRCh38
rs994439769 Chr.15: 42775701-42775701 on Build GRCh38
rs775186980 Chr.15: 42775593-42775593 on Build GRCh38
rs199715074 Chr.15: 42775433-42775433 on Build GRCh38
rs369226210 Chr.15: 42775612-42775612 on Build GRCh38
rs973549405 Chr.15: 42775656-42775656 on Build GRCh38
rs750560702 Chr.15: 42775526-42775526 on Build GRCh38
rs778872399 Chr.15: 42775563-42775563 on Build GRCh38
rs777438034 Chr.15: 42775539-42775539 on Build GRCh38
rs56039839 Chr.15: 42775634-42775634 on Build GRCh38
rs1000045139 Chr.15: 42775419-42775419 on Build GRCh38
rs866705172 Chr.15: 42775257-42775257 on Build GRCh38
rs756629999 Chr.15: 42775203-42775203 on Build GRCh38
rs376681856 Chr.15: 42775291-42775291 on Build GRCh38
rs910340635 Chr.15: 42775601-42775601 on Build GRCh38
rs904250448 Chr.15: 42775400-42775400 on Build GRCh38
rs778877514 Chr.15: 42775697-42775697 on Build GRCh38
rs375236448 Chr.15: 42775266-42775266 on Build GRCh38
rs200124857 Chr.15: 42775578-42775578 on Build GRCh38
rs1050680063 Chr.15: 42775594-42775594 on Build GRCh38
rs754442146 Chr.15: 42775440-42775440 on Build GRCh38
rs758714670 Chr.15: 42775387-42775387 on Build GRCh38
rs941755031 Chr.15: 42775624-42775624 on Build GRCh38
rs756437771 Chr.15: 42775318-42775318 on Build GRCh38
rs374669500 Chr.15: 42775462-42775462 on Build GRCh38
rs201633321 Chr.15: 42775436-42775436 on Build GRCh38
rs749178642 Chr.15: 42775422-42775422 on Build GRCh38
rs368873147 Chr.15: 42775472-42775472 on Build GRCh38
rs747789846 Chr.15: 42775396-42775398 on Build GRCh38
rs375368402 Chr.15: 42775369-42775369 on Build GRCh38
rs990190832 Chr.15: 42775510-42775510 on Build GRCh38
rs375035687 Chr.15: 42775580-42775580 on Build GRCh38
rs370977106 Chr.15: 42775236-42775236 on Build GRCh38
rs747901093 Chr.15: 42775263-42775263 on Build GRCh38
rs528648733 Chr.15: 42775230-42775230 on Build GRCh38
rs372204873 Chr.15: 42775476-42775476 on Build GRCh38

View Full Product Details