Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41748209-41748797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603898 c.132T>A p.Y44* Chr.4: 41748479-41748479 on Build GRCh38
5884023 c.119G>A p.S40N Chr.4: 41748492-41748492 on Build GRCh38
6984847 c.86C>T p.A29V Chr.4: 41748525-41748525 on Build GRCh38
262923 c.206C>A p.T69N Chr.4: 41748405-41748405 on Build GRCh38
4714164 c.52A>G p.M18V Chr.4: 41748559-41748559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756206203 Chr.4: 41748329-41748329 on Build GRCh38
rs763699416 Chr.4: 41748425-41748425 on Build GRCh38
rs773969259 Chr.4: 41748344-41748344 on Build GRCh38
rs1025677545 Chr.4: 41748255-41748255 on Build GRCh38
rs1047152254 Chr.4: 41748764-41748764 on Build GRCh38
rs908839332 Chr.4: 41748722-41748722 on Build GRCh38
rs968453180 Chr.4: 41748559-41748559 on Build GRCh38
rs921713677 Chr.4: 41748231-41748231 on Build GRCh38
rs554684781 Chr.4: 41748627-41748627 on Build GRCh38
rs766500239 Chr.4: 41748497-41748497 on Build GRCh38
rs750938350 Chr.4: 41748485-41748485 on Build GRCh38
rs532711949 Chr.4: 41748384-41748384 on Build GRCh38
rs750520429 Chr.4: 41748327-41748327 on Build GRCh38
rs373063040 Chr.4: 41748638-41748638 on Build GRCh38
rs759671110 Chr.4: 41748648-41748648 on Build GRCh38
rs953105370 Chr.4: 41748704-41748704 on Build GRCh38
rs745833730 Chr.4: 41748554-41748554 on Build GRCh38
rs755290304 Chr.4: 41748506-41748506 on Build GRCh38
rs761275637 Chr.4: 41748635-41748635 on Build GRCh38
rs781178952 Chr.4: 41748323-41748323 on Build GRCh38
rs775101470 Chr.4: 41748569-41748569 on Build GRCh38
rs376060053 Chr.4: 41748380-41748380 on Build GRCh38
rs550472167 Chr.4: 41748373-41748373 on Build GRCh38
rs374160638 Chr.4: 41748500-41748500 on Build GRCh38
rs746765437 Chr.4: 41748348-41748348 on Build GRCh38
rs769573717 Chr.4: 41748566-41748566 on Build GRCh38
rs538445545 Chr.4: 41748715-41748715 on Build GRCh38
rs922121255 Chr.4: 41748680-41748680 on Build GRCh38
rs201073251 Chr.4: 41748355-41748355 on Build GRCh38
rs577333579 Chr.4: 41748577-41748577 on Build GRCh38
rs779251110 Chr.4: 41748512-41748512 on Build GRCh38
rs754399397 Chr.4: 41748344-41748344 on Build GRCh38
rs781024969 Chr.4: 41748398-41748399 on Build GRCh38
rs878854616 Chr.4: 41748476-41748476 on Build GRCh38
rs534756790 Chr.4: 41748617-41748617 on Build GRCh38
rs371419084 Chr.4: 41748332-41748332 on Build GRCh38
rs951341615 Chr.4: 41748260-41748260 on Build GRCh38
rs879410848 Chr.4: 41748322-41748322 on Build GRCh38
rs776614949 Chr.4: 41748364-41748364 on Build GRCh38
rs774739194 Chr.4: 41748439-41748439 on Build GRCh38
rs756519209 Chr.4: 41748491-41748491 on Build GRCh38
rs1005823397 Chr.4: 41748306-41748306 on Build GRCh38
rs1040240739 Chr.4: 41748695-41748695 on Build GRCh38
rs910430385 Chr.4: 41748539-41748539 on Build GRCh38
rs946362319 Chr.4: 41748429-41748429 on Build GRCh38
rs748767112 Chr.4: 41748572-41748572 on Build GRCh38
rs200971068 Chr.4: 41748541-41748541 on Build GRCh38
rs762144117 Chr.4: 41748446-41748446 on Build GRCh38
rs749536907 Chr.4: 41748337-41748337 on Build GRCh38
rs908884850 Chr.4: 41748249-41748249 on Build GRCh38
rs755202332 Chr.4: 41748340-41748340 on Build GRCh38
rs559227588 Chr.4: 41748465-41748465 on Build GRCh38
rs773982008 Chr.4: 41748578-41748578 on Build GRCh38
rs980925920 Chr.4: 41748349-41748349 on Build GRCh38
rs887117495 Chr.4: 41748769-41748769 on Build GRCh38
rs746854983 Chr.4: 41748533-41748533 on Build GRCh38
rs567155526 Chr.4: 41748242-41748242 on Build GRCh38
rs200038327 Chr.4: 41748521-41748521 on Build GRCh38
rs534528291 Chr.4: 41748328-41748328 on Build GRCh38
rs73810366 Chr.4: 41748377-41748377 on Build GRCh38
rs587778605 Chr.4: 41748552-41748552 on Build GRCh38
rs561455489 Chr.4: 41748321-41748321 on Build GRCh38
rs111690228 Chr.4: 41748636-41748636 on Build GRCh38
rs969971317 Chr.4: 41748345-41748345 on Build GRCh38
rs780786684 Chr.4: 41748550-41748550 on Build GRCh38
rs1012348354 Chr.4: 41748458-41748458 on Build GRCh38
rs928566779 Chr.4: 41748771-41748771 on Build GRCh38
rs548642509 Chr.4: 41748232-41748232 on Build GRCh38
rs769914584 Chr.4: 41748731-41748731 on Build GRCh38
rs931630497 Chr.4: 41748234-41748234 on Build GRCh38
rs1043632230 Chr.4: 41748514-41748514 on Build GRCh38
rs1036276109 Chr.4: 41748767-41748767 on Build GRCh38
rs754017596 Chr.4: 41748647-41748647 on Build GRCh38

View Full Product Details