Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12649791-12650349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3891563 c.2226C>T p.F742F Chr.19: 12649951-12649951 on Build GRCh38
1390608 c.2192G>A p.R731H Chr.19: 12649985-12649985 on Build GRCh38
2816118 c.2146G>T p.G716W Chr.19: 12650120-12650120 on Build GRCh38
7234689 c.2202A>T p.T734T Chr.19: 12649975-12649975 on Build GRCh38
417751 c.2120G>A p.R707Q Chr.19: 12650146-12650146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373539727 Chr.19: 12649865-12649865 on Build GRCh38
rs3815914 Chr.19: 12649850-12649850 on Build GRCh38
rs761096537 Chr.19: 12650243-12650243 on Build GRCh38
rs373800486 Chr.19: 12650078-12650078 on Build GRCh38
rs142210875 Chr.19: 12649999-12649999 on Build GRCh38
rs543944689 Chr.19: 12650324-12650325 on Build GRCh38
rs747176828 Chr.19: 12649916-12649916 on Build GRCh38
rs754017897 Chr.19: 12650185-12650185 on Build GRCh38
rs569077441 Chr.19: 12649810-12649810 on Build GRCh38
rs764431833 Chr.19: 12650248-12650248 on Build GRCh38
rs755197711 Chr.19: 12649998-12649998 on Build GRCh38
rs533770473 Chr.19: 12649887-12649887 on Build GRCh38
rs778053346 Chr.19: 12650083-12650083 on Build GRCh38
rs751919256 Chr.19: 12650056-12650056 on Build GRCh38
rs770976179 Chr.19: 12650014-12650014 on Build GRCh38
rs377717440 Chr.19: 12649884-12649884 on Build GRCh38
rs745641762 Chr.19: 12650023-12650023 on Build GRCh38
rs373240866 Chr.19: 12649931-12649931 on Build GRCh38
rs781197939 Chr.19: 12649927-12649927 on Build GRCh38
rs749183690 Chr.19: 12650013-12650013 on Build GRCh38
rs939754783 Chr.19: 12650109-12650109 on Build GRCh38
rs746776449 Chr.19: 12650222-12650222 on Build GRCh38
rs761293269 Chr.19: 12650144-12650144 on Build GRCh38
rs80338680 Chr.19: 12649932-12649932 on Build GRCh38
rs747691435 Chr.19: 12650233-12650233 on Build GRCh38
rs61234887 Chr.19: 12649959-12649959 on Build GRCh38
rs988476350 Chr.19: 12649826-12649826 on Build GRCh38
rs1003984277 Chr.19: 12649860-12649860 on Build GRCh38
rs370551216 Chr.19: 12649955-12649955 on Build GRCh38
rs762877000 Chr.19: 12649933-12649933 on Build GRCh38
rs766154921 Chr.19: 12649880-12649880 on Build GRCh38
rs750833439 Chr.19: 12649904-12649904 on Build GRCh38
rs779966472 Chr.19: 12650215-12650215 on Build GRCh38
rs757769028 Chr.19: 12649891-12649891 on Build GRCh38
rs143829874 Chr.19: 12650009-12650009 on Build GRCh38
rs138817592 Chr.19: 12649985-12649985 on Build GRCh38
rs778745390 Chr.19: 12650204-12650204 on Build GRCh38
rs940088931 Chr.19: 12650239-12650239 on Build GRCh38
rs150533763 Chr.19: 12650147-12650147 on Build GRCh38
rs769957397 Chr.19: 12650003-12650003 on Build GRCh38
rs550622289 Chr.19: 12650262-12650262 on Build GRCh38
rs1057517166 Chr.19: 12650223-12650223 on Build GRCh38
rs376804252 Chr.19: 12649990-12649990 on Build GRCh38
rs754071126 Chr.19: 12650257-12650257 on Build GRCh38
rs776158673 Chr.19: 12650124-12650124 on Build GRCh38
rs141965228 Chr.19: 12650123-12650123 on Build GRCh38
rs761715309 Chr.19: 12649868-12649868 on Build GRCh38
rs988974694 Chr.19: 12650165-12650165 on Build GRCh38
rs1022927322 Chr.19: 12649912-12649912 on Build GRCh38
rs955787667 Chr.19: 12650125-12650125 on Build GRCh38
rs769767974 Chr.19: 12649869-12649869 on Build GRCh38
rs775010162 Chr.19: 12650116-12650116 on Build GRCh38
rs921831751 Chr.19: 12650217-12650217 on Build GRCh38
rs28639634 Chr.19: 12649910-12649910 on Build GRCh38
rs555203186 Chr.19: 12649896-12649896 on Build GRCh38
rs368551873 Chr.19: 12650276-12650276 on Build GRCh38
rs770786458 Chr.19: 12649953-12649953 on Build GRCh38
rs561720564 Chr.19: 12650168-12650168 on Build GRCh38
rs201380558 Chr.19: 12650050-12650050 on Build GRCh38
rs761987850 Chr.19: 12650260-12650260 on Build GRCh38
rs201978883 Chr.19: 12650084-12650084 on Build GRCh38
rs140449678 Chr.19: 12650172-12650172 on Build GRCh38
rs760151408 Chr.19: 12650235-12650235 on Build GRCh38
rs368669177 Chr.19: 12650032-12650032 on Build GRCh38
rs374501850 Chr.19: 12649881-12649881 on Build GRCh38
rs750890285 Chr.19: 12649972-12649972 on Build GRCh38
rs867109973 Chr.19: 12650117-12650117 on Build GRCh38
rs764376758 Chr.19: 12650181-12650181 on Build GRCh38
rs745749915 Chr.19: 12650212-12650212 on Build GRCh38
rs1035913927 Chr.19: 12650218-12650218 on Build GRCh38
rs746606611 Chr.19: 12650118-12650118 on Build GRCh38
rs371812948 Chr.19: 12650146-12650146 on Build GRCh38
rs199700264 Chr.19: 12649986-12649986 on Build GRCh38
rs768347546 Chr.19: 12650226-12650226 on Build GRCh38
rs376110556 Chr.19: 12650193-12650193 on Build GRCh38
rs773182870 Chr.19: 12649872-12649872 on Build GRCh38
rs766842420 Chr.19: 12650053-12650053 on Build GRCh38
rs772448821 Chr.19: 12650237-12650237 on Build GRCh38
rs759094537 Chr.19: 12649885-12649885 on Build GRCh38
rs367602761 Chr.19: 12650101-12650101 on Build GRCh38
rs974658068 Chr.19: 12650196-12650196 on Build GRCh38
rs756279141 Chr.19: 12650080-12650080 on Build GRCh38
rs864621993 Chr.19: 12650129-12650129 on Build GRCh38
rs765753479 Chr.19: 12649966-12649966 on Build GRCh38
rs864621987 Chr.19: 12649946-12649946 on Build GRCh38
rs767667453 Chr.19: 12650076-12650076 on Build GRCh38
rs544632771 Chr.19: 12649827-12649827 on Build GRCh38
rs987043625 Chr.19: 12650323-12650323 on Build GRCh38
rs763526229 Chr.19: 12650042-12650042 on Build GRCh38
rs886044335 Chr.19: 12649905-12649906 on Build GRCh38
rs774156075 Chr.19: 12649954-12649954 on Build GRCh38
rs201318291 Chr.19: 12650000-12650000 on Build GRCh38
rs778385288 Chr.19: 12650107-12650107 on Build GRCh38
rs141212446 Chr.19: 12649920-12649920 on Build GRCh38
rs142754988 Chr.19: 12650214-12650214 on Build GRCh38
rs761646956 Chr.19: 12649888-12649889 on Build GRCh38
rs757361100 Chr.19: 12650190-12650190 on Build GRCh38
rs572289342 Chr.19: 12649904-12649905 on Build GRCh38
rs1037527582 Chr.19: 12650258-12650258 on Build GRCh38
rs774350579 Chr.19: 12650022-12650022 on Build GRCh38
rs148134639 Chr.19: 12650108-12650108 on Build GRCh38
rs767192781 Chr.19: 12649956-12649956 on Build GRCh38
rs771858783 Chr.19: 12650110-12650110 on Build GRCh38
rs759826825 Chr.19: 12650164-12650164 on Build GRCh38
rs80338679 Chr.19: 12650103-12650103 on Build GRCh38
rs954278875 Chr.19: 12650245-12650245 on Build GRCh38
rs770253455 Chr.19: 12650060-12650060 on Build GRCh38
rs776107165 Chr.19: 12650048-12650048 on Build GRCh38
rs762822402 Chr.19: 12649877-12649877 on Build GRCh38
rs529363964 Chr.19: 12650241-12650241 on Build GRCh38
rs750179985 Chr.19: 12649940-12649943 on Build GRCh38
rs760618868 Chr.19: 12650189-12650191 on Build GRCh38
rs200424566 Chr.19: 12650227-12650227 on Build GRCh38
rs752234339 Chr.19: 12649886-12649886 on Build GRCh38
rs775158789 Chr.19: 12650028-12650028 on Build GRCh38
rs769284524 Chr.19: 12650234-12650234 on Build GRCh38
rs771668348 Chr.19: 12650026-12650026 on Build GRCh38
rs778884658 Chr.19: 12650104-12650104 on Build GRCh38
rs186573686 Chr.19: 12649832-12649832 on Build GRCh38

View Full Product Details