Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27548244-27548799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245731 c.1262A>T p.Q421L Chr.9: 27548420-27548420 on Build GRCh38
3848298 c.1386T>C p.F462F Chr.9: 27548296-27548296 on Build GRCh38
7112823 c.1315T>G p.L439V Chr.9: 27548367-27548367 on Build GRCh38
1108018 c.1254C>T p.D418D Chr.9: 27548562-27548562 on Build GRCh38
3745959 c.1408A>G p.T470A Chr.9: 27548274-27548274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370093286 Chr.9: 27548462-27548462 on Build GRCh38
rs878962858 Chr.9: 27548488-27548488 on Build GRCh38
rs60460283 Chr.9: 27548772-27548772 on Build GRCh38
rs774493813 Chr.9: 27548353-27548353 on Build GRCh38
rs769477910 Chr.9: 27548558-27548558 on Build GRCh38
rs766148523 Chr.9: 27548465-27548468 on Build GRCh38
rs373026414 Chr.9: 27548435-27548436 on Build GRCh38
rs917836310 Chr.9: 27548379-27548379 on Build GRCh38
rs766107319 Chr.9: 27548386-27548386 on Build GRCh38
rs753432326 Chr.9: 27548275-27548275 on Build GRCh38
rs771894999 Chr.9: 27548537-27548537 on Build GRCh38
rs780934681 Chr.9: 27548628-27548628 on Build GRCh38
rs756267679 Chr.9: 27548415-27548415 on Build GRCh38
rs747091754 Chr.9: 27548688-27548689 on Build GRCh38
rs369969754 Chr.9: 27548682-27548682 on Build GRCh38
rs898227726 Chr.9: 27548466-27548466 on Build GRCh38
rs886063844 Chr.9: 27548436-27548436 on Build GRCh38
rs775400975 Chr.9: 27548336-27548336 on Build GRCh38
rs748290291 Chr.9: 27548532-27548532 on Build GRCh38
rs779161252 Chr.9: 27548527-27548527 on Build GRCh38
rs13285738 Chr.9: 27548461-27548461 on Build GRCh38
rs374772194 Chr.9: 27548672-27548672 on Build GRCh38
rs762570472 Chr.9: 27548412-27548413 on Build GRCh38
rs182259442 Chr.9: 27548549-27548549 on Build GRCh38
rs761335050 Chr.9: 27548442-27548443 on Build GRCh38
rs762581307 Chr.9: 27548568-27548568 on Build GRCh38
rs749166700 Chr.9: 27548433-27548434 on Build GRCh38
rs926457792 Chr.9: 27548340-27548340 on Build GRCh38
rs931007664 Chr.9: 27548724-27548724 on Build GRCh38
rs376801304 Chr.9: 27548679-27548679 on Build GRCh38
rs752075581 Chr.9: 27548398-27548398 on Build GRCh38
rs755859608 Chr.9: 27548430-27548430 on Build GRCh38
rs149274966 Chr.9: 27548307-27548307 on Build GRCh38
rs772868096 Chr.9: 27548380-27548380 on Build GRCh38
rs750237326 Chr.9: 27548429-27548435 on Build GRCh38
rs554657570 Chr.9: 27548485-27548485 on Build GRCh38
rs747372712 Chr.9: 27548295-27548295 on Build GRCh38
rs754838983 Chr.9: 27548542-27548544 on Build GRCh38
rs530261789 Chr.9: 27548586-27548586 on Build GRCh38
rs368371587 Chr.9: 27548385-27548385 on Build GRCh38
rs755916502 Chr.9: 27548462-27548468 on Build GRCh38
rs889659172 Chr.9: 27548759-27548759 on Build GRCh38
rs753837506 Chr.9: 27548507-27548508 on Build GRCh38
rs368145554 Chr.9: 27548523-27548523 on Build GRCh38
rs772249544 Chr.9: 27548434-27548435 on Build GRCh38
rs1049489429 Chr.9: 27548734-27548734 on Build GRCh38
rs991962885 Chr.9: 27548656-27548656 on Build GRCh38
rs748114242 Chr.9: 27548424-27548424 on Build GRCh38
rs377349627 Chr.9: 27548773-27548773 on Build GRCh38
rs376653879 Chr.9: 27548458-27548462 on Build GRCh38
rs201593885 Chr.9: 27548462-27548462 on Build GRCh38
rs115623999 Chr.9: 27548538-27548538 on Build GRCh38
rs763702959 Chr.9: 27548440-27548441 on Build GRCh38
rs376835503 Chr.9: 27548715-27548715 on Build GRCh38
rs767652287 Chr.9: 27548439-27548440 on Build GRCh38
rs373632829 Chr.9: 27548732-27548733 on Build GRCh38
rs185358772 Chr.9: 27548632-27548632 on Build GRCh38
rs778862260 Chr.9: 27548617-27548618 on Build GRCh38
rs761630312 Chr.9: 27548370-27548370 on Build GRCh38
rs768119232 Chr.9: 27548472-27548472 on Build GRCh38
rs774370838 Chr.9: 27548421-27548421 on Build GRCh38
rs1037213590 Chr.9: 27548674-27548674 on Build GRCh38
rs760887239 Chr.9: 27548578-27548578 on Build GRCh38
rs369166616 Chr.9: 27548289-27548289 on Build GRCh38
rs375040006 Chr.9: 27548332-27548332 on Build GRCh38
rs573486774 Chr.9: 27548499-27548499 on Build GRCh38
rs919590950 Chr.9: 27548717-27548717 on Build GRCh38
rs759984347 Chr.9: 27548436-27548440 on Build GRCh38
rs755216584 Chr.9: 27548587-27548587 on Build GRCh38
rs199877251 Chr.9: 27548687-27548687 on Build GRCh38
rs779339076 Chr.9: 27548591-27548591 on Build GRCh38
rs1049972951 Chr.9: 27548495-27548495 on Build GRCh38
rs968170641 Chr.9: 27548313-27548313 on Build GRCh38
rs200703028 Chr.9: 27548397-27548397 on Build GRCh38
rs535961666 Chr.9: 27548471-27548471 on Build GRCh38
rs886063843 Chr.9: 27548435-27548437 on Build GRCh38
rs754552300 Chr.9: 27548391-27548391 on Build GRCh38
rs371626518 Chr.9: 27548459-27548462 on Build GRCh38
rs546365564 Chr.9: 27548766-27548766 on Build GRCh38
rs752022465 Chr.9: 27548443-27548443 on Build GRCh38
rs768858247 Chr.9: 27548420-27548420 on Build GRCh38
rs902104051 Chr.9: 27548528-27548528 on Build GRCh38
rs748177144 Chr.9: 27548436-27548439 on Build GRCh38
rs2305045 Chr.9: 27548514-27548514 on Build GRCh38
rs954039651 Chr.9: 27548329-27548329 on Build GRCh38
rs773709157 Chr.9: 27548441-27548442 on Build GRCh38
rs767226139 Chr.9: 27548373-27548373 on Build GRCh38
rs2643806 Chr.9: 27548639-27548639 on Build GRCh38
rs776218894 Chr.9: 27548556-27548556 on Build GRCh38
rs770228283 Chr.9: 27548610-27548610 on Build GRCh38
rs780257522 Chr.9: 27548438-27548439 on Build GRCh38
rs569786818 Chr.9: 27548318-27548318 on Build GRCh38
rs757569780 Chr.9: 27548404-27548404 on Build GRCh38
rs866566387 Chr.9: 27548764-27548764 on Build GRCh38
rs746100459 Chr.9: 27548310-27548310 on Build GRCh38
rs1036670830 Chr.9: 27548460-27548460 on Build GRCh38
rs2484318 Chr.9: 27548425-27548425 on Build GRCh38
rs201734930 Chr.9: 27548562-27548562 on Build GRCh38
rs190477376 Chr.9: 27548406-27548406 on Build GRCh38
rs11292923 Chr.9: 27548461-27548461 on Build GRCh38
rs779971360 Chr.9: 27548430-27548435 on Build GRCh38
rs931060537 Chr.9: 27548482-27548482 on Build GRCh38
rs377048084 Chr.9: 27548461-27548462 on Build GRCh38
rs753188234 Chr.9: 27548296-27548297 on Build GRCh38
rs771083562 Chr.9: 27548436-27548441 on Build GRCh38
rs753335393 Chr.9: 27548390-27548390 on Build GRCh38
rs928103463 Chr.9: 27548506-27548506 on Build GRCh38
rs814031 Chr.9: 27548721-27548721 on Build GRCh38
rs765877989 Chr.9: 27548272-27548272 on Build GRCh38
rs751296187 Chr.9: 27548424-27548424 on Build GRCh38
rs749427374 Chr.9: 27548345-27548345 on Build GRCh38
rs746990973 Chr.9: 27548625-27548625 on Build GRCh38
rs939774089 Chr.9: 27548659-27548659 on Build GRCh38
rs966030940 Chr.9: 27548271-27548271 on Build GRCh38
rs200572381 Chr.9: 27548535-27548535 on Build GRCh38
rs747306049 Chr.9: 27548436-27548437 on Build GRCh38
rs754124108 Chr.9: 27548519-27548519 on Build GRCh38
rs759218702 Chr.9: 27548557-27548557 on Build GRCh38
rs369558106 Chr.9: 27548716-27548716 on Build GRCh38
rs201564924 Chr.9: 27548518-27548518 on Build GRCh38
rs749761510 Chr.9: 27548708-27548708 on Build GRCh38
rs541965231 Chr.9: 27548606-27548606 on Build GRCh38
rs781087590 Chr.9: 27548428-27548435 on Build GRCh38
rs572661170 Chr.9: 27548436-27548438 on Build GRCh38
rs374582800 Chr.9: 27548460-27548462 on Build GRCh38
rs758512117 Chr.9: 27548609-27548609 on Build GRCh38
rs756831457 Chr.9: 27548427-27548435 on Build GRCh38
rs886063842 Chr.9: 27548435-27548436 on Build GRCh38
rs199587552 Chr.9: 27548433-27548433 on Build GRCh38
rs812272 Chr.9: 27548419-27548419 on Build GRCh38
rs201585198 Chr.9: 27548712-27548712 on Build GRCh38
rs750942348 Chr.9: 27548577-27548577 on Build GRCh38
rs34602100 Chr.9: 27548583-27548583 on Build GRCh38
rs552524653 Chr.9: 27548751-27548751 on Build GRCh38
rs773541376 Chr.9: 27548695-27548695 on Build GRCh38
rs811641 Chr.9: 27548612-27548612 on Build GRCh38
rs1088056 Chr.9: 27548323-27548323 on Build GRCh38
rs768698995 Chr.9: 27548352-27548352 on Build GRCh38
rs767903441 Chr.9: 27548571-27548571 on Build GRCh38
rs780380295 Chr.9: 27548416-27548416 on Build GRCh38
rs775188943 Chr.9: 27548643-27548643 on Build GRCh38
rs371785983 Chr.9: 27548431-27548431 on Build GRCh38
rs376385318 Chr.9: 27548382-27548382 on Build GRCh38
rs757625044 Chr.9: 27548302-27548302 on Build GRCh38
rs764751687 Chr.9: 27548436-27548436 on Build GRCh38
rs1011255904 Chr.9: 27548763-27548763 on Build GRCh38
rs144786050 Chr.9: 27548771-27548771 on Build GRCh38
rs767103403 Chr.9: 27548454-27548462 on Build GRCh38
rs141063383 Chr.9: 27548278-27548278 on Build GRCh38
rs2453563 Chr.9: 27548407-27548407 on Build GRCh38
rs745710266 Chr.9: 27548636-27548636 on Build GRCh38
rs201135029 Chr.9: 27548430-27548430 on Build GRCh38

View Full Product Details