Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27556477-27556812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029957 c.979A>T p.N327Y Chr.9: 27556673-27556673 on Build GRCh38
1108022 c.876G>A p.V292V Chr.9: 27556776-27556776 on Build GRCh38
6044948 c.1055C>T p.T352M Chr.9: 27556597-27556597 on Build GRCh38
6115560 c.951G>A p.Q317Q Chr.9: 27556701-27556701 on Build GRCh38
3699644 c.985C>T p.R329C Chr.9: 27556667-27556667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370472336 Chr.9: 27556765-27556765 on Build GRCh38
rs1026481197 Chr.9: 27556611-27556611 on Build GRCh38
rs370756199 Chr.9: 27556670-27556670 on Build GRCh38
rs753735631 Chr.9: 27556681-27556681 on Build GRCh38
rs757944597 Chr.9: 27556632-27556632 on Build GRCh38
rs141784061 Chr.9: 27556651-27556651 on Build GRCh38
rs758281611 Chr.9: 27556516-27556516 on Build GRCh38
rs770353129 Chr.9: 27556611-27556612 on Build GRCh38
rs777232451 Chr.9: 27556552-27556552 on Build GRCh38
rs201818484 Chr.9: 27556541-27556541 on Build GRCh38
rs762221739 Chr.9: 27556525-27556525 on Build GRCh38
rs754395436 Chr.9: 27556700-27556700 on Build GRCh38
rs773251849 Chr.9: 27556667-27556667 on Build GRCh38
rs763285533 Chr.9: 27556575-27556575 on Build GRCh38
rs910769995 Chr.9: 27556695-27556695 on Build GRCh38
rs139502204 Chr.9: 27556714-27556714 on Build GRCh38
rs371377376 Chr.9: 27556634-27556634 on Build GRCh38
rs374369837 Chr.9: 27556672-27556672 on Build GRCh38
rs747599155 Chr.9: 27556534-27556534 on Build GRCh38
rs752423770 Chr.9: 27556626-27556626 on Build GRCh38
rs759330431 Chr.9: 27556692-27556692 on Build GRCh38
rs753650999 Chr.9: 27556602-27556602 on Build GRCh38
rs576983400 Chr.9: 27556610-27556610 on Build GRCh38
rs746409630 Chr.9: 27556557-27556557 on Build GRCh38
rs963419379 Chr.9: 27556592-27556592 on Build GRCh38
rs10122902 Chr.9: 27556782-27556782 on Build GRCh38
rs35726666 Chr.9: 27556756-27556756 on Build GRCh38
rs539512959 Chr.9: 27556675-27556675 on Build GRCh38
rs146043466 Chr.9: 27556658-27556658 on Build GRCh38
rs764976547 Chr.9: 27556697-27556697 on Build GRCh38
rs989795030 Chr.9: 27556535-27556535 on Build GRCh38
rs777628989 Chr.9: 27556710-27556710 on Build GRCh38
rs972582676 Chr.9: 27556598-27556598 on Build GRCh38
rs746077661 Chr.9: 27556540-27556541 on Build GRCh38
rs769024683 Chr.9: 27556760-27556760 on Build GRCh38
rs764371434 Chr.9: 27556577-27556577 on Build GRCh38
rs201961571 Chr.9: 27556666-27556666 on Build GRCh38
rs779927591 Chr.9: 27556519-27556519 on Build GRCh38
rs373419325 Chr.9: 27556637-27556637 on Build GRCh38
rs770465523 Chr.9: 27556558-27556558 on Build GRCh38
rs746566978 Chr.9: 27556642-27556642 on Build GRCh38
rs750317119 Chr.9: 27556591-27556591 on Build GRCh38
rs774506585 Chr.9: 27556656-27556656 on Build GRCh38
rs948461835 Chr.9: 27556554-27556554 on Build GRCh38
rs780494226 Chr.9: 27556650-27556650 on Build GRCh38
rs556983155 Chr.9: 27556588-27556588 on Build GRCh38
rs201261393 Chr.9: 27556778-27556778 on Build GRCh38
rs557931293 Chr.9: 27556597-27556597 on Build GRCh38
rs771724358 Chr.9: 27556537-27556537 on Build GRCh38
rs774440971 Chr.9: 27556580-27556580 on Build GRCh38
rs766352238 Chr.9: 27556586-27556586 on Build GRCh38
rs754633522 Chr.9: 27556520-27556520 on Build GRCh38
rs775901504 Chr.9: 27556574-27556574 on Build GRCh38
rs758101832 Chr.9: 27556701-27556701 on Build GRCh38
rs767605056 Chr.9: 27556581-27556581 on Build GRCh38
rs754736903 Chr.9: 27556603-27556603 on Build GRCh38
rs568888933 Chr.9: 27556590-27556591 on Build GRCh38
rs538012382 Chr.9: 27556662-27556662 on Build GRCh38
rs766277022 Chr.9: 27556679-27556679 on Build GRCh38
rs780733794 Chr.9: 27556751-27556751 on Build GRCh38
rs1013494001 Chr.9: 27556504-27556504 on Build GRCh38
rs770323658 Chr.9: 27556649-27556649 on Build GRCh38
rs756724177 Chr.9: 27556730-27556730 on Build GRCh38

View Full Product Details