Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55146664-55146798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5407712 c.537C>T p.D179D Chr.7: 55146718-55146718 on Build GRCh38
252489 c.493C>T p.R165W Chr.7: 55146674-55146674 on Build GRCh38
6950556 c.530C>T p.S177L Chr.7: 55146711-55146711 on Build GRCh38
5407715 c.537C>T p.D179D Chr.7: 55146718-55146718 on Build GRCh38
4384114 c.523A>G p.N175D Chr.7: 55146704-55146704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749002473 Chr.7: 55146706-55146706 on Build GRCh38
rs772071414 Chr.7: 55146728-55146728 on Build GRCh38
rs921852102 Chr.7: 55146732-55146732 on Build GRCh38
rs770290445 Chr.7: 55146711-55146711 on Build GRCh38
rs372921874 Chr.7: 55146757-55146757 on Build GRCh38
rs974428039 Chr.7: 55146721-55146721 on Build GRCh38
rs1032993509 Chr.7: 55146746-55146746 on Build GRCh38
rs758945260 Chr.7: 55146690-55146690 on Build GRCh38
rs771969559 Chr.7: 55146760-55146760 on Build GRCh38
rs745755245 Chr.7: 55146715-55146715 on Build GRCh38
rs762104816 Chr.7: 55146756-55146756 on Build GRCh38
rs375037710 Chr.7: 55146739-55146739 on Build GRCh38
rs17289686 Chr.7: 55146694-55146694 on Build GRCh38
rs17336437 Chr.7: 55146712-55146712 on Build GRCh38
rs7801956 Chr.7: 55146750-55146750 on Build GRCh38
rs185023939 Chr.7: 55146749-55146749 on Build GRCh38
rs967951017 Chr.7: 55146748-55146748 on Build GRCh38
rs777401847 Chr.7: 55146700-55146700 on Build GRCh38
rs776979310 Chr.7: 55146753-55146753 on Build GRCh38
rs530692924 Chr.7: 55146683-55146683 on Build GRCh38

View Full Product Details