Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41745666-41745834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7383808 c.925G>C p.V309L Chr.4: 41745827-41745827 on Build GRCh38
5449994 c.919G>A p.A307T Chr.4: 41745833-41745833 on Build GRCh38
2847560 c.920C>G p.A307G Chr.4: 41745832-41745832 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs895428574 Chr.4: 41745742-41745761 on Build GRCh38
rs955624967 Chr.4: 41745750-41745750 on Build GRCh38
rs774951337 Chr.4: 41745774-41745775 on Build GRCh38
rs987470852 Chr.4: 41745754-41745754 on Build GRCh38
rs771616235 Chr.4: 41745775-41745783 on Build GRCh38
rs973793027 Chr.4: 41745775-41745780 on Build GRCh38
rs999505638 Chr.4: 41745689-41745689 on Build GRCh38
rs754671744 Chr.4: 41745769-41745769 on Build GRCh38
rs770841700 Chr.4: 41745788-41745788 on Build GRCh38
rs780019991 Chr.4: 41745766-41745766 on Build GRCh38
rs759556353 Chr.4: 41745790-41745790 on Build GRCh38
rs905041383 Chr.4: 41745703-41745703 on Build GRCh38
rs768798757 Chr.4: 41745784-41745784 on Build GRCh38
rs779171868 Chr.4: 41745787-41745787 on Build GRCh38
rs749346904 Chr.4: 41745779-41745779 on Build GRCh38
rs578163582 Chr.4: 41745753-41745753 on Build GRCh38
rs751212535 Chr.4: 41745810-41745810 on Build GRCh38
rs763704619 Chr.4: 41745802-41745802 on Build GRCh38
rs886059413 Chr.4: 41745749-41745749 on Build GRCh38
rs75913938 Chr.4: 41745727-41745727 on Build GRCh38
rs538080267 Chr.4: 41745733-41745733 on Build GRCh38
rs989877179 Chr.4: 41745738-41745738 on Build GRCh38
rs763380864 Chr.4: 41745775-41745777 on Build GRCh38
rs957517040 Chr.4: 41745729-41745729 on Build GRCh38
rs769726959 Chr.4: 41745793-41745793 on Build GRCh38
rs762497253 Chr.4: 41745801-41745801 on Build GRCh38
rs756546457 Chr.4: 41745765-41745765 on Build GRCh38
rs774897796 Chr.4: 41745798-41745798 on Build GRCh38
rs558416040 Chr.4: 41745747-41745747 on Build GRCh38
rs911357344 Chr.4: 41745772-41745772 on Build GRCh38

View Full Product Details