Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55155792-55155933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6954371 c.954G>A p.M318I Chr.7: 55155894-55155894 on Build GRCh38
6981709 c.951G>C p.E317D Chr.7: 55155891-55155891 on Build GRCh38
6962238 c.898_900delGTG p.V301delV Chr.7: 55155838-55155840 on Build GRCh38
6458157 c.940G>A p.D314N Chr.7: 55155880-55155880 on Build GRCh38
1600351 c.977G>C p.C326S Chr.7: 55155917-55155917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182002674 Chr.7: 55155852-55155852 on Build GRCh38
rs772053702 Chr.7: 55155885-55155885 on Build GRCh38
rs767820212 Chr.7: 55155814-55155814 on Build GRCh38
rs745658347 Chr.7: 55155884-55155884 on Build GRCh38
rs753038774 Chr.7: 55155815-55155815 on Build GRCh38
rs770879879 Chr.7: 55155875-55155875 on Build GRCh38
rs757810509 Chr.7: 55155826-55155826 on Build GRCh38
rs776791214 Chr.7: 55155904-55155904 on Build GRCh38
rs202182545 Chr.7: 55155858-55155858 on Build GRCh38
rs201040971 Chr.7: 55155909-55155909 on Build GRCh38
rs367680488 Chr.7: 55155908-55155908 on Build GRCh38
rs552062864 Chr.7: 55155880-55155880 on Build GRCh38
rs149321481 Chr.7: 55155878-55155878 on Build GRCh38
rs17289893 Chr.7: 55155861-55155861 on Build GRCh38
rs761844164 Chr.7: 55155907-55155907 on Build GRCh38
rs760826609 Chr.7: 55155819-55155819 on Build GRCh38
rs779094647 Chr.7: 55155841-55155841 on Build GRCh38
rs529223915 Chr.7: 55155821-55155821 on Build GRCh38
rs764038041 Chr.7: 55155903-55155903 on Build GRCh38
rs566001525 Chr.7: 55155855-55155855 on Build GRCh38
rs749132706 Chr.7: 55155862-55155862 on Build GRCh38
rs760984499 Chr.7: 55155900-55155900 on Build GRCh38
rs775800262 Chr.7: 55155894-55155894 on Build GRCh38
rs754229667 Chr.7: 55155825-55155825 on Build GRCh38
rs758748662 Chr.7: 55155911-55155911 on Build GRCh38

View Full Product Details