Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100628090-100628331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4766884 c.1466G>A p.W489* Chr.7: 100628231-100628231 on Build GRCh38
6800597 c.1414_1416delCTC p.L472delL Chr.7: 100628281-100628283 on Build GRCh38
4908565 c.1403G>A p.R468H Chr.7: 100628294-100628294 on Build GRCh38
4994575 c.1460C>T p.T487M Chr.7: 100628237-100628237 on Build GRCh38
341459 c.1401C>A p.P467P Chr.7: 100628296-100628296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762935630 Chr.7: 100628229-100628229 on Build GRCh38
rs533880980 Chr.7: 100628247-100628247 on Build GRCh38
rs144148322 Chr.7: 100628226-100628226 on Build GRCh38
rs200201634 Chr.7: 100628211-100628211 on Build GRCh38
rs201082151 Chr.7: 100628209-100628209 on Build GRCh38
rs374113312 Chr.7: 100628210-100628210 on Build GRCh38
rs748905972 Chr.7: 100628264-100628264 on Build GRCh38
rs377542243 Chr.7: 100628188-100628188 on Build GRCh38
rs1040713439 Chr.7: 100628206-100628206 on Build GRCh38
rs772672626 Chr.7: 100628168-100628168 on Build GRCh38
rs759247993 Chr.7: 100628180-100628180 on Build GRCh38
rs747728512 Chr.7: 100628154-100628154 on Build GRCh38
rs747312334 Chr.7: 100628143-100628143 on Build GRCh38
rs780410371 Chr.7: 100628200-100628200 on Build GRCh38
rs41295899 Chr.7: 100628248-100628248 on Build GRCh38
rs374546301 Chr.7: 100628214-100628214 on Build GRCh38
rs758992254 Chr.7: 100628197-100628197 on Build GRCh38
rs779079099 Chr.7: 100628121-100628121 on Build GRCh38
rs186378421 Chr.7: 100628208-100628208 on Build GRCh38
rs770754691 Chr.7: 100628173-100628173 on Build GRCh38
rs202038214 Chr.7: 100628277-100628277 on Build GRCh38
rs931476373 Chr.7: 100628137-100628137 on Build GRCh38
rs767913945 Chr.7: 100628241-100628241 on Build GRCh38
rs778438972 Chr.7: 100628266-100628266 on Build GRCh38
rs373415641 Chr.7: 100628125-100628125 on Build GRCh38
rs766437336 Chr.7: 100628111-100628111 on Build GRCh38
rs769302506 Chr.7: 100628161-100628161 on Build GRCh38
rs145499236 Chr.7: 100628300-100628300 on Build GRCh38
rs774161209 Chr.7: 100628212-100628212 on Build GRCh38
rs376230804 Chr.7: 100628207-100628207 on Build GRCh38
rs756733318 Chr.7: 100628265-100628265 on Build GRCh38
rs80338886 Chr.7: 100628228-100628228 on Build GRCh38
rs762922296 Chr.7: 100628171-100628171 on Build GRCh38
rs770202105 Chr.7: 100628295-100628295 on Build GRCh38
rs766412097 Chr.7: 100628236-100628236 on Build GRCh38
rs767276889 Chr.7: 100628183-100628183 on Build GRCh38
rs890069610 Chr.7: 100628179-100628179 on Build GRCh38
rs773308636 Chr.7: 100628312-100628312 on Build GRCh38
rs759461055 Chr.7: 100628216-100628216 on Build GRCh38
rs772014889 Chr.7: 100628217-100628217 on Build GRCh38
rs139178017 Chr.7: 100628224-100628224 on Build GRCh38
rs373025420 Chr.7: 100628205-100628205 on Build GRCh38
rs768409892 Chr.7: 100628145-100628145 on Build GRCh38
rs773320398 Chr.7: 100628307-100628307 on Build GRCh38
rs765439073 Chr.7: 100628191-100628191 on Build GRCh38
rs777401563 Chr.7: 100628259-100628259 on Build GRCh38
rs775385253 Chr.7: 100628222-100628222 on Build GRCh38
rs780814385 Chr.7: 100628148-100628148 on Build GRCh38
rs752025865 Chr.7: 100628203-100628203 on Build GRCh38
rs750499411 Chr.7: 100628192-100628192 on Build GRCh38
rs80338885 Chr.7: 100628294-100628294 on Build GRCh38
rs770286722 Chr.7: 100628195-100628195 on Build GRCh38
rs771822800 Chr.7: 100628213-100628213 on Build GRCh38
rs987977631 Chr.7: 100628115-100628115 on Build GRCh38
rs146530042 Chr.7: 100628237-100628237 on Build GRCh38
rs774262230 Chr.7: 100628174-100628174 on Build GRCh38
rs773050231 Chr.7: 100628299-100628299 on Build GRCh38
rs772104483 Chr.7: 100628288-100628288 on Build GRCh38
rs1049922942 Chr.7: 100628159-100628159 on Build GRCh38
rs777532178 Chr.7: 100628124-100628124 on Build GRCh38
rs754126409 Chr.7: 100628185-100628185 on Build GRCh38

View Full Product Details