Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 872763-873036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522086 c.1456G>A p.G486S Chr.20: 873016-873016 on Build GRCh38
6094016 c.1446C>A p.H482Q Chr.20: 873026-873026 on Build GRCh38
3405301 c.1438C>T p.R480C Chr.20: 873034-873034 on Build GRCh38
1713055 c.1496G>A p.R499Q Chr.20: 872976-872976 on Build GRCh38
6347697 c.1485C>A p.R495R Chr.20: 872987-872987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754971249 Chr.20: 872958-872958 on Build GRCh38
rs68006621 Chr.20: 872791-872791 on Build GRCh38
rs370917173 Chr.20: 872941-872941 on Build GRCh38
rs764259122 Chr.20: 872923-872923 on Build GRCh38
rs758172223 Chr.20: 872977-872977 on Build GRCh38
rs893102560 Chr.20: 872835-872835 on Build GRCh38
rs541775009 Chr.20: 872949-872949 on Build GRCh38
rs1888087 Chr.20: 872792-872792 on Build GRCh38
rs368678351 Chr.20: 872919-872919 on Build GRCh38
rs770310800 Chr.20: 872996-872996 on Build GRCh38
rs750130199 Chr.20: 872906-872906 on Build GRCh38
rs370362896 Chr.20: 872792-872792 on Build GRCh38
rs201271383 Chr.20: 872991-872991 on Build GRCh38
rs909870451 Chr.20: 872823-872823 on Build GRCh38
rs1010240188 Chr.20: 872854-872854 on Build GRCh38
rs182043253 Chr.20: 872994-872994 on Build GRCh38
rs1001724421 Chr.20: 872798-872798 on Build GRCh38
rs769116790 Chr.20: 873008-873008 on Build GRCh38
rs541080353 Chr.20: 872995-872995 on Build GRCh38
rs368267857 Chr.20: 872989-872989 on Build GRCh38
rs556018104 Chr.20: 872891-872891 on Build GRCh38
rs541864923 Chr.20: 872791-872792 on Build GRCh38
rs374332736 Chr.20: 872948-872948 on Build GRCh38
rs906081082 Chr.20: 872797-872797 on Build GRCh38
rs866219144 Chr.20: 872869-872869 on Build GRCh38
rs200751369 Chr.20: 872910-872910 on Build GRCh38
rs751850679 Chr.20: 872938-872938 on Build GRCh38
rs974366515 Chr.20: 872908-872908 on Build GRCh38
rs775833469 Chr.20: 872918-872918 on Build GRCh38
rs539495797 Chr.20: 872866-872866 on Build GRCh38
rs753697482 Chr.20: 872999-872999 on Build GRCh38
rs747977524 Chr.20: 872972-872972 on Build GRCh38
rs770474502 Chr.20: 872988-872988 on Build GRCh38
rs1054744418 Chr.20: 872825-872825 on Build GRCh38
rs750528347 Chr.20: 872954-872954 on Build GRCh38
rs11907018 Chr.20: 872859-872859 on Build GRCh38
rs866650260 Chr.20: 872783-872783 on Build GRCh38
rs746720846 Chr.20: 872981-872981 on Build GRCh38
rs754898686 Chr.20: 873015-873015 on Build GRCh38
rs28504506 Chr.20: 872959-872959 on Build GRCh38
rs1008352804 Chr.20: 872867-872867 on Build GRCh38
rs775939876 Chr.20: 872997-872997 on Build GRCh38

View Full Product Details