Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81705410-81705683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1488699 c.1913G>A p.G638E Chr.7: 81705487-81705487 on Build GRCh38
1623130 c.1984G>T p.E662* Chr.7: 81705416-81705416 on Build GRCh38
6515099 c.1876G>T p.D626Y Chr.7: 81705524-81705524 on Build GRCh38
6908827 c.1926C>A p.C642* Chr.7: 81705474-81705474 on Build GRCh38
6983159 c.1912G>A p.G638R Chr.7: 81705488-81705488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376395424 Chr.7: 81705534-81705534 on Build GRCh38
rs747052903 Chr.7: 81705604-81705604 on Build GRCh38
rs747158538 Chr.7: 81705511-81705511 on Build GRCh38
rs764695573 Chr.7: 81705552-81705552 on Build GRCh38
rs746110664 Chr.7: 81705502-81705502 on Build GRCh38
rs780359807 Chr.7: 81705602-81705602 on Build GRCh38
rs200374420 Chr.7: 81705654-81705654 on Build GRCh38
rs540676769 Chr.7: 81705542-81705542 on Build GRCh38
rs757640030 Chr.7: 81705560-81705560 on Build GRCh38
rs369418750 Chr.7: 81705456-81705456 on Build GRCh38
rs769920904 Chr.7: 81705579-81705582 on Build GRCh38
rs954312187 Chr.7: 81705522-81705522 on Build GRCh38
rs770672389 Chr.7: 81705649-81705649 on Build GRCh38
rs145494248 Chr.7: 81705509-81705509 on Build GRCh38
rs750636675 Chr.7: 81705566-81705566 on Build GRCh38
rs184717721 Chr.7: 81705561-81705561 on Build GRCh38
rs755575139 Chr.7: 81705531-81705531 on Build GRCh38
rs150109563 Chr.7: 81705458-81705458 on Build GRCh38
rs878880281 Chr.7: 81705597-81705597 on Build GRCh38
rs926889688 Chr.7: 81705545-81705545 on Build GRCh38
rs756602776 Chr.7: 81705481-81705481 on Build GRCh38
rs373545882 Chr.7: 81705528-81705528 on Build GRCh38
rs748539273 Chr.7: 81705593-81705593 on Build GRCh38
rs766007676 Chr.7: 81705435-81705435 on Build GRCh38
rs545249104 Chr.7: 81705461-81705461 on Build GRCh38
rs565056780 Chr.7: 81705496-81705496 on Build GRCh38
rs769585774 Chr.7: 81705512-81705512 on Build GRCh38
rs202215700 Chr.7: 81705466-81705466 on Build GRCh38
rs779267373 Chr.7: 81705500-81705500 on Build GRCh38
rs931329793 Chr.7: 81705622-81705622 on Build GRCh38
rs777627774 Chr.7: 81705635-81705635 on Build GRCh38
rs778395987 Chr.7: 81705483-81705483 on Build GRCh38
rs758523692 Chr.7: 81705582-81705582 on Build GRCh38
rs751270979 Chr.7: 81705446-81705446 on Build GRCh38
rs774153258 Chr.7: 81705652-81705652 on Build GRCh38
rs13235690 Chr.7: 81705626-81705626 on Build GRCh38
rs770723689 Chr.7: 81705520-81705520 on Build GRCh38
rs754226152 Chr.7: 81705494-81705494 on Build GRCh38
rs771594786 Chr.7: 81705658-81705658 on Build GRCh38
rs754457294 Chr.7: 81705557-81705557 on Build GRCh38
rs749226169 Chr.7: 81705637-81705637 on Build GRCh38
rs773198515 Chr.7: 81705515-81705515 on Build GRCh38
rs866398503 Chr.7: 81705645-81705645 on Build GRCh38
rs755141326 Chr.7: 81705619-81705619 on Build GRCh38
rs371826587 Chr.7: 81705533-81705533 on Build GRCh38
rs1012056012 Chr.7: 81705448-81705448 on Build GRCh38
rs13238580 Chr.7: 81705627-81705627 on Build GRCh38
rs762721825 Chr.7: 81705516-81705516 on Build GRCh38
rs779495509 Chr.7: 81705547-81705547 on Build GRCh38

View Full Product Details