Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16988093-16988336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750611 c.2658G>A p.S886S Chr.1: 16988207-16988207 on Build GRCh38
899714 c.2789C>T p.S930L Chr.1: 16988208-16988208 on Build GRCh38
4484165 c.2777C>T p.S926F Chr.1: 16988220-16988220 on Build GRCh38
5881107 c.2631G>C p.R877S Chr.1: 16988234-16988234 on Build GRCh38
3802888 c.2709C>T p.S903S Chr.1: 16988156-16988156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983312062 Chr.1: 16988164-16988164 on Build GRCh38
rs755542233 Chr.1: 16988201-16988201 on Build GRCh38
rs796448884 Chr.1: 16988291-16988293 on Build GRCh38
rs576215702 Chr.1: 16988262-16988262 on Build GRCh38
rs922495404 Chr.1: 16988199-16988199 on Build GRCh38
rs752015774 Chr.1: 16988278-16988278 on Build GRCh38
rs373413809 Chr.1: 16988149-16988149 on Build GRCh38
rs765505590 Chr.1: 16988259-16988259 on Build GRCh38
rs754030102 Chr.1: 16988156-16988156 on Build GRCh38
rs371934965 Chr.1: 16988260-16988260 on Build GRCh38
rs367638044 Chr.1: 16988251-16988251 on Build GRCh38
rs375014566 Chr.1: 16988192-16988192 on Build GRCh38
rs368404440 Chr.1: 16988198-16988198 on Build GRCh38
rs779656439 Chr.1: 16988303-16988303 on Build GRCh38
rs886045578 Chr.1: 16988189-16988189 on Build GRCh38
rs148721722 Chr.1: 16988139-16988139 on Build GRCh38
rs775423066 Chr.1: 16988231-16988231 on Build GRCh38
rs762012587 Chr.1: 16988255-16988255 on Build GRCh38
rs755414129 Chr.1: 16988281-16988281 on Build GRCh38
rs144557304 Chr.1: 16988138-16988138 on Build GRCh38
rs758709320 Chr.1: 16988195-16988195 on Build GRCh38
rs114407349 Chr.1: 16988250-16988250 on Build GRCh38
rs745504274 Chr.1: 16988299-16988299 on Build GRCh38
rs536168405 Chr.1: 16988130-16988130 on Build GRCh38
rs370384911 Chr.1: 16988118-16988118 on Build GRCh38
rs12129596 Chr.1: 16988202-16988202 on Build GRCh38
rs55708915 Chr.1: 16988161-16988161 on Build GRCh38
rs760518104 Chr.1: 16988239-16988239 on Build GRCh38
rs560897844 Chr.1: 16988301-16988301 on Build GRCh38
rs200702121 Chr.1: 16988284-16988284 on Build GRCh38
rs746804111 Chr.1: 16988304-16988304 on Build GRCh38
rs567866687 Chr.1: 16988227-16988227 on Build GRCh38
rs141447403 Chr.1: 16988293-16988293 on Build GRCh38
rs143237568 Chr.1: 16988272-16988272 on Build GRCh38
rs370356637 Chr.1: 16988200-16988200 on Build GRCh38
rs148336170 Chr.1: 16988291-16988291 on Build GRCh38
rs3738815 Chr.1: 16988207-16988207 on Build GRCh38
rs763274429 Chr.1: 16988268-16988268 on Build GRCh38
rs764264628 Chr.1: 16988155-16988155 on Build GRCh38
rs778562581 Chr.1: 16988208-16988208 on Build GRCh38
rs759283607 Chr.1: 16988117-16988117 on Build GRCh38
rs201437622 Chr.1: 16988296-16988296 on Build GRCh38
rs765560804 Chr.1: 16988177-16988177 on Build GRCh38
rs564643512 Chr.1: 16988226-16988226 on Build GRCh38
rs755438239 Chr.1: 16988228-16988228 on Build GRCh38

View Full Product Details