Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1362603-1362846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672464 c.611G>A p.G204D Chr.16: 1362612-1362612 on Build GRCh38
4058425 c.751G>T p.E251* Chr.16: 1362834-1362834 on Build GRCh38
1375957 c.646G>A p.A216T Chr.16: 1362647-1362647 on Build GRCh38
2143663 c.699T>C p.G233G Chr.16: 1362700-1362700 on Build GRCh38
434663 c.630G>A p.R210R Chr.16: 1362631-1362631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748861038 Chr.16: 1362680-1362680 on Build GRCh38
rs202129898 Chr.16: 1362739-1362739 on Build GRCh38
rs776277092 Chr.16: 1362783-1362784 on Build GRCh38
rs753661059 Chr.16: 1362766-1362766 on Build GRCh38
rs368747296 Chr.16: 1362730-1362730 on Build GRCh38
rs761527949 Chr.16: 1362644-1362644 on Build GRCh38
rs150965148 Chr.16: 1362685-1362685 on Build GRCh38
rs966758681 Chr.16: 1362682-1362682 on Build GRCh38
rs371060844 Chr.16: 1362748-1362748 on Build GRCh38
rs141035347 Chr.16: 1362742-1362742 on Build GRCh38
rs775527344 Chr.16: 1362679-1362679 on Build GRCh38
rs532275192 Chr.16: 1362703-1362703 on Build GRCh38
rs756093689 Chr.16: 1362671-1362671 on Build GRCh38
rs893287710 Chr.16: 1362786-1362786 on Build GRCh38
rs201844106 Chr.16: 1362652-1362652 on Build GRCh38
rs757938105 Chr.16: 1362793-1362793 on Build GRCh38
rs202080062 Chr.16: 1362714-1362714 on Build GRCh38
rs772287534 Chr.16: 1362684-1362684 on Build GRCh38
rs990613310 Chr.16: 1362702-1362702 on Build GRCh38
rs193302858 Chr.16: 1362640-1362640 on Build GRCh38
rs142548931 Chr.16: 1362666-1362666 on Build GRCh38
rs775816256 Chr.16: 1362701-1362701 on Build GRCh38
rs777120261 Chr.16: 1362648-1362648 on Build GRCh38
rs555595061 Chr.16: 1362810-1362810 on Build GRCh38
rs778631467 Chr.16: 1362738-1362738 on Build GRCh38
rs969101852 Chr.16: 1362759-1362760 on Build GRCh38
rs138332265 Chr.16: 1362785-1362785 on Build GRCh38
rs761341825 Chr.16: 1362797-1362798 on Build GRCh38
rs200239398 Chr.16: 1362770-1362770 on Build GRCh38
rs774431331 Chr.16: 1362753-1362753 on Build GRCh38
rs201263537 Chr.16: 1362747-1362747 on Build GRCh38
rs768377103 Chr.16: 1362642-1362642 on Build GRCh38
rs780289639 Chr.16: 1362806-1362806 on Build GRCh38
rs751817263 Chr.16: 1362664-1362664 on Build GRCh38
rs925124634 Chr.16: 1362633-1362633 on Build GRCh38
rs199607037 Chr.16: 1362752-1362752 on Build GRCh38
rs761837002 Chr.16: 1362754-1362754 on Build GRCh38
rs764405882 Chr.16: 1362661-1362661 on Build GRCh38
rs748692260 Chr.16: 1362641-1362641 on Build GRCh38
rs1054675795 Chr.16: 1362783-1362783 on Build GRCh38
rs754489307 Chr.16: 1362684-1362684 on Build GRCh38
rs780930809 Chr.16: 1362628-1362628 on Build GRCh38
rs762888658 Chr.16: 1362658-1362658 on Build GRCh38
rs761243739 Chr.16: 1362752-1362753 on Build GRCh38
rs368890072 Chr.16: 1362755-1362755 on Build GRCh38
rs776107971 Chr.16: 1362712-1362712 on Build GRCh38
rs747594923 Chr.16: 1362741-1362741 on Build GRCh38
rs762029401 Chr.16: 1362706-1362706 on Build GRCh38
rs764994151 Chr.16: 1362810-1362811 on Build GRCh38
rs757003667 Chr.16: 1362624-1362624 on Build GRCh38
rs200094858 Chr.16: 1362691-1362691 on Build GRCh38
rs746271025 Chr.16: 1362636-1362637 on Build GRCh38
rs750155751 Chr.16: 1362668-1362668 on Build GRCh38
rs890055206 Chr.16: 1362631-1362631 on Build GRCh38
rs774108877 Chr.16: 1362643-1362643 on Build GRCh38
rs771844889 Chr.16: 1362692-1362692 on Build GRCh38
rs201392900 Chr.16: 1362750-1362750 on Build GRCh38
rs746160240 Chr.16: 1362697-1362697 on Build GRCh38
rs774413019 Chr.16: 1362704-1362704 on Build GRCh38
rs779807567 Chr.16: 1362672-1362672 on Build GRCh38
rs768131774 Chr.16: 1362639-1362639 on Build GRCh38
rs573130947 Chr.16: 1362771-1362771 on Build GRCh38
rs766167977 Chr.16: 1362715-1362715 on Build GRCh38
rs766147394 Chr.16: 1362756-1362756 on Build GRCh38
rs756918285 Chr.16: 1362800-1362800 on Build GRCh38
rs771470862 Chr.16: 1362645-1362645 on Build GRCh38
rs754928775 Chr.16: 1362731-1362731 on Build GRCh38
rs552248197 Chr.16: 1362638-1362638 on Build GRCh38
rs777140384 Chr.16: 1362694-1362694 on Build GRCh38
rs911440995 Chr.16: 1362758-1362758 on Build GRCh38
rs112925416 Chr.16: 1362632-1362632 on Build GRCh38
rs779356410 Chr.16: 1362818-1362818 on Build GRCh38
rs751216882 Chr.16: 1362798-1362798 on Build GRCh38
rs137853827 Chr.16: 1362689-1362689 on Build GRCh38
rs749610702 Chr.16: 1362809-1362809 on Build GRCh38
rs763757057 Chr.16: 1362774-1362775 on Build GRCh38
rs374564369 Chr.16: 1362698-1362698 on Build GRCh38
rs760155537 Chr.16: 1362650-1362650 on Build GRCh38
rs762827756 Chr.16: 1362713-1362713 on Build GRCh38
rs1001955796 Chr.16: 1362764-1362764 on Build GRCh38

View Full Product Details