Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49200084-49200357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561379 c.340G>T p.D114Y Chr.11: 49200326-49200326 on Build GRCh38
4821648 c.327G>A p.E109E Chr.11: 49200339-49200339 on Build GRCh38
927691 c.316G>A p.D106N Chr.11: 49200350-49200350 on Build GRCh38
4546667 c.404G>A p.G135E Chr.11: 49200262-49200262 on Build GRCh38
3998431 c.333A>T p.A111A Chr.11: 49200333-49200333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034310110 Chr.11: 49200171-49200171 on Build GRCh38
rs760249223 Chr.11: 49200255-49200255 on Build GRCh38
rs767438602 Chr.11: 49200312-49200312 on Build GRCh38
rs202680 Chr.11: 49200333-49200333 on Build GRCh38
rs567592268 Chr.11: 49200231-49200234 on Build GRCh38
rs990158372 Chr.11: 49200194-49200194 on Build GRCh38
rs958687982 Chr.11: 49200192-49200192 on Build GRCh38
rs766804724 Chr.11: 49200298-49200298 on Build GRCh38
rs931614121 Chr.11: 49200290-49200290 on Build GRCh38
rs369440863 Chr.11: 49200304-49200304 on Build GRCh38
rs186234267 Chr.11: 49200190-49200190 on Build GRCh38
rs7128652 Chr.11: 49200271-49200271 on Build GRCh38
rs565277046 Chr.11: 49200228-49200228 on Build GRCh38
rs376750461 Chr.11: 49200166-49200166 on Build GRCh38
rs971310416 Chr.11: 49200150-49200150 on Build GRCh38
rs752820178 Chr.11: 49200332-49200332 on Build GRCh38
rs1015189889 Chr.11: 49200146-49200146 on Build GRCh38
rs763279653 Chr.11: 49200279-49200279 on Build GRCh38
rs887534665 Chr.11: 49200114-49200114 on Build GRCh38
rs563140296 Chr.11: 49200233-49200233 on Build GRCh38
rs914622828 Chr.11: 49200199-49200199 on Build GRCh38
rs267602927 Chr.11: 49200263-49200263 on Build GRCh38
rs1004633288 Chr.11: 49200130-49200130 on Build GRCh38
rs377142674 Chr.11: 49200284-49200284 on Build GRCh38
rs532027840 Chr.11: 49200246-49200246 on Build GRCh38
rs763728006 Chr.11: 49200262-49200262 on Build GRCh38
rs977559594 Chr.11: 49200230-49200231 on Build GRCh38
rs745852202 Chr.11: 49200241-49200241 on Build GRCh38
rs546200035 Chr.11: 49200229-49200229 on Build GRCh38
rs771437533 Chr.11: 49200244-49200244 on Build GRCh38
rs751970048 Chr.11: 49200303-49200303 on Build GRCh38
rs772477133 Chr.11: 49200246-49200247 on Build GRCh38

View Full Product Details