Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74920350-74920595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6532557 c.450C>G p.C150W Chr.17: 74920386-74920386 on Build GRCh38
1231916 c.424G>A p.E142K Chr.17: 74920412-74920412 on Build GRCh38
4547934 c.434G>T p.R145L Chr.17: 74920402-74920402 on Build GRCh38
1231914 c.440T>A p.I147N Chr.17: 74920396-74920396 on Build GRCh38
6817887 c.430G>T p.E144* Chr.17: 74920406-74920406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995800790 Chr.17: 74920433-74920433 on Build GRCh38
rs774827536 Chr.17: 74920570-74920570 on Build GRCh38
rs751735275 Chr.17: 74920524-74920524 on Build GRCh38
rs760465755 Chr.17: 74920493-74920493 on Build GRCh38
rs765050197 Chr.17: 74920517-74920517 on Build GRCh38
rs200082225 Chr.17: 74920403-74920403 on Build GRCh38
rs954934463 Chr.17: 74920462-74920462 on Build GRCh38
rs140951373 Chr.17: 74920447-74920447 on Build GRCh38
rs748592728 Chr.17: 74920425-74920425 on Build GRCh38
rs756461951 Chr.17: 74920541-74920541 on Build GRCh38
rs375077331 Chr.17: 74920451-74920451 on Build GRCh38
rs370887019 Chr.17: 74920431-74920431 on Build GRCh38
rs778163212 Chr.17: 74920544-74920544 on Build GRCh38
rs775117744 Chr.17: 74920492-74920492 on Build GRCh38
rs1010293771 Chr.17: 74920393-74920393 on Build GRCh38
rs751196707 Chr.17: 74920368-74920368 on Build GRCh38
rs748325007 Chr.17: 74920537-74920537 on Build GRCh38
rs745450960 Chr.17: 74920572-74920572 on Build GRCh38
rs920379501 Chr.17: 74920547-74920547 on Build GRCh38
rs202005955 Chr.17: 74920402-74920402 on Build GRCh38
rs773610870 Chr.17: 74920441-74920441 on Build GRCh38
rs773698651 Chr.17: 74920380-74920380 on Build GRCh38
rs963261823 Chr.17: 74920554-74920554 on Build GRCh38
rs368407264 Chr.17: 74920574-74920574 on Build GRCh38
rs759026779 Chr.17: 74920457-74920457 on Build GRCh38
rs947865484 Chr.17: 74920553-74920553 on Build GRCh38
rs759149374 Chr.17: 74920373-74920373 on Build GRCh38
rs763891710 Chr.17: 74920496-74920496 on Build GRCh38
rs771517713 Chr.17: 74920556-74920556 on Build GRCh38
rs770131812 Chr.17: 74920427-74920427 on Build GRCh38
rs747261903 Chr.17: 74920415-74920415 on Build GRCh38
rs757174476 Chr.17: 74920516-74920516 on Build GRCh38
rs141791769 Chr.17: 74920400-74920400 on Build GRCh38
rs561119699 Chr.17: 74920476-74920476 on Build GRCh38
rs149529031 Chr.17: 74920526-74920526 on Build GRCh38
rs144686462 Chr.17: 74920575-74920575 on Build GRCh38
rs111033465 Chr.17: 74920448-74920448 on Build GRCh38
rs374260443 Chr.17: 74920536-74920536 on Build GRCh38
rs781540158 Chr.17: 74920418-74920418 on Build GRCh38
rs755288377 Chr.17: 74920417-74920417 on Build GRCh38
rs749586302 Chr.17: 74920555-74920555 on Build GRCh38
rs111033466 Chr.17: 74920412-74920412 on Build GRCh38
rs753740036 Chr.17: 74920505-74920505 on Build GRCh38
rs1021385697 Chr.17: 74920380-74920380 on Build GRCh38
rs587776546 Chr.17: 74920441-74920442 on Build GRCh38
rs1021745794 Chr.17: 74920407-74920407 on Build GRCh38
rs886053390 Chr.17: 74920488-74920488 on Build GRCh38
rs201822644 Chr.17: 74920377-74920377 on Build GRCh38

View Full Product Details