Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100630820-100631093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
393746 c.1264G>A p.E422K Chr.7: 100630895-100630895 on Build GRCh38
3684371 c.1249A>G p.I417V Chr.7: 100630910-100630910 on Build GRCh38
6231250 c.1140G>A p.W380* Chr.7: 100631019-100631019 on Build GRCh38
3949579 c.1234A>C p.N412H Chr.7: 100630925-100630925 on Build GRCh38
7393642 c.1138T>A p.W380R Chr.7: 100631021-100631021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757050571 Chr.7: 100631010-100631010 on Build GRCh38
rs755112053 Chr.7: 100631012-100631012 on Build GRCh38
rs766014113 Chr.7: 100630854-100630854 on Build GRCh38
rs370005636 Chr.7: 100630919-100630919 on Build GRCh38
rs999797341 Chr.7: 100631006-100631006 on Build GRCh38
rs778433259 Chr.7: 100631030-100631030 on Build GRCh38
rs781371035 Chr.7: 100631021-100631021 on Build GRCh38
rs760716258 Chr.7: 100631005-100631005 on Build GRCh38
rs958164035 Chr.7: 100630972-100630972 on Build GRCh38
rs774163818 Chr.7: 100630998-100630998 on Build GRCh38
rs752704464 Chr.7: 100630954-100630954 on Build GRCh38
rs150883560 Chr.7: 100630917-100630917 on Build GRCh38
rs777383473 Chr.7: 100630878-100630878 on Build GRCh38
rs888964552 Chr.7: 100631070-100631070 on Build GRCh38
rs1050275436 Chr.7: 100631067-100631067 on Build GRCh38
rs202022975 Chr.7: 100630966-100630966 on Build GRCh38
rs771448341 Chr.7: 100631034-100631034 on Build GRCh38
rs749669683 Chr.7: 100630982-100630982 on Build GRCh38
rs779767961 Chr.7: 100630865-100630865 on Build GRCh38
rs577055819 Chr.7: 100631043-100631043 on Build GRCh38
rs142005608 Chr.7: 100630896-100630896 on Build GRCh38
rs149590949 Chr.7: 100630980-100630980 on Build GRCh38
rs868455004 Chr.7: 100630899-100630899 on Build GRCh38
rs756739081 Chr.7: 100631027-100631027 on Build GRCh38
rs751801070 Chr.7: 100630850-100630852 on Build GRCh38
rs185470250 Chr.7: 100630873-100630873 on Build GRCh38
rs143185818 Chr.7: 100630916-100630916 on Build GRCh38
rs371880627 Chr.7: 100630957-100630957 on Build GRCh38
rs762643498 Chr.7: 100630849-100630849 on Build GRCh38
rs202221581 Chr.7: 100631041-100631041 on Build GRCh38
rs766104011 Chr.7: 100630945-100630945 on Build GRCh38
rs866804730 Chr.7: 100631004-100631012 on Build GRCh38
rs146901123 Chr.7: 100630900-100630900 on Build GRCh38
rs767086840 Chr.7: 100631002-100631002 on Build GRCh38
rs767495840 Chr.7: 100631012-100631013 on Build GRCh38
rs773757652 Chr.7: 100630981-100630981 on Build GRCh38
rs748150579 Chr.7: 100631025-100631025 on Build GRCh38
rs557506122 Chr.7: 100630979-100630979 on Build GRCh38
rs773669738 Chr.7: 100630936-100630936 on Build GRCh38
rs751828156 Chr.7: 100631011-100631011 on Build GRCh38
rs762231662 Chr.7: 100630840-100630840 on Build GRCh38
rs141101651 Chr.7: 100630910-100630910 on Build GRCh38
rs753609418 Chr.7: 100630964-100630964 on Build GRCh38
rs759064628 Chr.7: 100631001-100631001 on Build GRCh38
rs41303495 Chr.7: 100631032-100631032 on Build GRCh38
rs762731224 Chr.7: 100630941-100630941 on Build GRCh38
rs200597967 Chr.7: 100630874-100630874 on Build GRCh38
rs201334869 Chr.7: 100630867-100630867 on Build GRCh38
rs139383159 Chr.7: 100630907-100630907 on Build GRCh38
rs764007668 Chr.7: 100631008-100631008 on Build GRCh38
rs753747243 Chr.7: 100631027-100631027 on Build GRCh38
rs144917130 Chr.7: 100630953-100630953 on Build GRCh38
rs375150921 Chr.7: 100630955-100630955 on Build GRCh38
rs80338882 Chr.7: 100630973-100630973 on Build GRCh38
rs138189378 Chr.7: 100630967-100630967 on Build GRCh38
rs80338883 Chr.7: 100630926-100630928 on Build GRCh38
rs764276305 Chr.7: 100630846-100630847 on Build GRCh38
rs547889468 Chr.7: 100630843-100630843 on Build GRCh38
rs771358868 Chr.7: 100630984-100630984 on Build GRCh38
rs945399761 Chr.7: 100630997-100630997 on Build GRCh38
rs774834320 Chr.7: 100631003-100631003 on Build GRCh38
rs747076627 Chr.7: 100630908-100630908 on Build GRCh38
rs751078792 Chr.7: 100630859-100630859 on Build GRCh38

View Full Product Details