Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50579799-50580068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361191 c.925C>G p.Q309E Chr.22: 50579976-50579976 on Build GRCh38
3414219 c.871G>A p.E291K Chr.22: 50580030-50580030 on Build GRCh38
4105235 c.865A>G p.T289A Chr.22: 50580036-50580036 on Build GRCh38
5934114 c.847G>A p.E283K Chr.22: 50580054-50580054 on Build GRCh38
7318468 c.919G>C p.E307Q Chr.22: 50579982-50579982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140020889 Chr.22: 50580031-50580031 on Build GRCh38
rs771040904 Chr.22: 50579841-50579841 on Build GRCh38
rs766044832 Chr.22: 50579827-50579827 on Build GRCh38
rs139059552 Chr.22: 50579976-50579976 on Build GRCh38
rs772869314 Chr.22: 50579874-50579874 on Build GRCh38
rs976125914 Chr.22: 50579885-50579885 on Build GRCh38
rs916899523 Chr.22: 50579995-50579995 on Build GRCh38
rs374030717 Chr.22: 50579949-50579949 on Build GRCh38
rs769598051 Chr.22: 50580002-50580002 on Build GRCh38
rs779131876 Chr.22: 50579985-50579985 on Build GRCh38
rs143061031 Chr.22: 50579993-50579993 on Build GRCh38
rs560351546 Chr.22: 50579858-50579858 on Build GRCh38
rs370202172 Chr.22: 50580008-50580008 on Build GRCh38
rs549171476 Chr.22: 50579964-50579964 on Build GRCh38
rs750097672 Chr.22: 50579819-50579819 on Build GRCh38
rs548666534 Chr.22: 50579898-50579898 on Build GRCh38
rs908462354 Chr.22: 50579892-50579892 on Build GRCh38
rs984131563 Chr.22: 50579925-50579925 on Build GRCh38
rs1017738897 Chr.22: 50579859-50579859 on Build GRCh38
rs387907069 Chr.22: 50579979-50579979 on Build GRCh38
rs751219415 Chr.22: 50579944-50579944 on Build GRCh38
rs774461264 Chr.22: 50579846-50579846 on Build GRCh38
rs749587921 Chr.22: 50579838-50579838 on Build GRCh38
rs766099951 Chr.22: 50579939-50579939 on Build GRCh38
rs757507890 Chr.22: 50579983-50579983 on Build GRCh38
rs778253283 Chr.22: 50579975-50579975 on Build GRCh38
rs775547378 Chr.22: 50579996-50579996 on Build GRCh38
rs147485527 Chr.22: 50579999-50579999 on Build GRCh38
rs369532290 Chr.22: 50579835-50579835 on Build GRCh38
rs762690035 Chr.22: 50580020-50580020 on Build GRCh38
rs752126398 Chr.22: 50579912-50579912 on Build GRCh38
rs983929252 Chr.22: 50579877-50579877 on Build GRCh38
rs777932969 Chr.22: 50579837-50579837 on Build GRCh38
rs767076133 Chr.22: 50579962-50579962 on Build GRCh38
rs188223105 Chr.22: 50579868-50579868 on Build GRCh38
rs766225706 Chr.22: 50580028-50580028 on Build GRCh38
rs376480861 Chr.22: 50579865-50579865 on Build GRCh38
rs370329322 Chr.22: 50579929-50579929 on Build GRCh38
rs752131662 Chr.22: 50580042-50580042 on Build GRCh38
rs200919604 Chr.22: 50579818-50579818 on Build GRCh38
rs374544105 Chr.22: 50580030-50580030 on Build GRCh38
rs749194510 Chr.22: 50579899-50579899 on Build GRCh38
rs751093137 Chr.22: 50579834-50579834 on Build GRCh38
rs765150933 Chr.22: 50579869-50579869 on Build GRCh38
rs373763595 Chr.22: 50579849-50579849 on Build GRCh38
rs772568756 Chr.22: 50579819-50579819 on Build GRCh38
rs375639229 Chr.22: 50579965-50579965 on Build GRCh38
rs992511759 Chr.22: 50579931-50579931 on Build GRCh38
rs757983603 Chr.22: 50579821-50579821 on Build GRCh38

View Full Product Details