Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1361653-1361908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4255909 c.211T>C p.C71R Chr.16: 1361775-1361775 on Build GRCh38
6672466 c.183C>T p.P61P Chr.16: 1361747-1361747 on Build GRCh38
5655139 c.196C>T p.R66* Chr.16: 1361760-1361760 on Build GRCh38
4058421 c.235T>C p.Y79H Chr.16: 1361873-1361873 on Build GRCh38
4475819 c.201C>G p.L67L Chr.16: 1361765-1361765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762446439 Chr.16: 1361732-1361732 on Build GRCh38
rs778178434 Chr.16: 1361889-1361889 on Build GRCh38
rs193302848 Chr.16: 1361760-1361760 on Build GRCh38
rs748167959 Chr.16: 1361704-1361704 on Build GRCh38
rs774835763 Chr.16: 1361783-1361783 on Build GRCh38
rs773452586 Chr.16: 1361876-1361881 on Build GRCh38
rs200915537 Chr.16: 1361852-1361852 on Build GRCh38
rs200362559 Chr.16: 1361805-1361805 on Build GRCh38
rs376327160 Chr.16: 1361867-1361867 on Build GRCh38
rs371413637 Chr.16: 1361826-1361827 on Build GRCh38
rs745356243 Chr.16: 1361831-1361831 on Build GRCh38
rs746507546 Chr.16: 1361715-1361715 on Build GRCh38
rs372039531 Chr.16: 1361774-1361774 on Build GRCh38
rs748941730 Chr.16: 1361881-1361881 on Build GRCh38
rs772637099 Chr.16: 1361795-1361795 on Build GRCh38
rs757318601 Chr.16: 1361869-1361869 on Build GRCh38
rs772275436 Chr.16: 1361840-1361840 on Build GRCh38
rs763430254 Chr.16: 1361725-1361725 on Build GRCh38
rs369333639 Chr.16: 1361864-1361864 on Build GRCh38
rs769814085 Chr.16: 1361844-1361845 on Build GRCh38
rs1017508203 Chr.16: 1361680-1361680 on Build GRCh38
rs973297006 Chr.16: 1361724-1361724 on Build GRCh38
rs762024309 Chr.16: 1361787-1361787 on Build GRCh38
rs751592131 Chr.16: 1361822-1361822 on Build GRCh38
rs112850896 Chr.16: 1361871-1361871 on Build GRCh38
rs754010312 Chr.16: 1361755-1361755 on Build GRCh38
rs771874727 Chr.16: 1361710-1361710 on Build GRCh38
rs200574942 Chr.16: 1361748-1361748 on Build GRCh38
rs748272191 Chr.16: 1361814-1361815 on Build GRCh38
rs769110491 Chr.16: 1361726-1361726 on Build GRCh38
rs758065510 Chr.16: 1361821-1361821 on Build GRCh38
rs376679416 Chr.16: 1361798-1361798 on Build GRCh38
rs201381376 Chr.16: 1361841-1361841 on Build GRCh38
rs745552342 Chr.16: 1361775-1361775 on Build GRCh38
rs759479821 Chr.16: 1361855-1361855 on Build GRCh38
rs779778381 Chr.16: 1361880-1361880 on Build GRCh38
rs754595108 Chr.16: 1361884-1361884 on Build GRCh38
rs988175540 Chr.16: 1361742-1361742 on Build GRCh38
rs199782632 Chr.16: 1361808-1361808 on Build GRCh38
rs555838948 Chr.16: 1361818-1361818 on Build GRCh38
rs371836175 Chr.16: 1361842-1361842 on Build GRCh38
rs776359831 Chr.16: 1361721-1361721 on Build GRCh38
rs749952316 Chr.16: 1361875-1361875 on Build GRCh38
rs752553225 Chr.16: 1361817-1361817 on Build GRCh38
rs775539912 Chr.16: 1361863-1361863 on Build GRCh38
rs751393135 Chr.16: 1361868-1361868 on Build GRCh38
rs774546219 Chr.16: 1361731-1361731 on Build GRCh38
rs368983807 Chr.16: 1361876-1361876 on Build GRCh38
rs757226714 Chr.16: 1361826-1361826 on Build GRCh38
rs569826327 Chr.16: 1361692-1361692 on Build GRCh38
rs552402857 Chr.16: 1361767-1361767 on Build GRCh38
rs376636252 Chr.16: 1361833-1361833 on Build GRCh38
rs780912876 Chr.16: 1361830-1361830 on Build GRCh38
rs201566045 Chr.16: 1361800-1361800 on Build GRCh38
rs373810840 Chr.16: 1361816-1361816 on Build GRCh38
rs771424681 Chr.16: 1361846-1361846 on Build GRCh38
rs777411480 Chr.16: 1361714-1361714 on Build GRCh38
rs778810991 Chr.16: 1361761-1361761 on Build GRCh38
rs200507633 Chr.16: 1361797-1361797 on Build GRCh38
rs144929410 Chr.16: 1361832-1361832 on Build GRCh38
rs767502955 Chr.16: 1361873-1361873 on Build GRCh38
rs777517766 Chr.16: 1361768-1361768 on Build GRCh38
rs140264422 Chr.16: 1361747-1361747 on Build GRCh38
rs769323972 Chr.16: 1361782-1361782 on Build GRCh38
rs767895135 Chr.16: 1361744-1361744 on Build GRCh38
rs979478155 Chr.16: 1361684-1361684 on Build GRCh38
rs952698680 Chr.16: 1361867-1361874 on Build GRCh38
rs754728320 Chr.16: 1361693-1361693 on Build GRCh38
rs748576549 Chr.16: 1361838-1361838 on Build GRCh38
rs9926432 Chr.16: 1361804-1361804 on Build GRCh38
rs372004052 Chr.16: 1361814-1361814 on Build GRCh38
rs766619396 Chr.16: 1361750-1361750 on Build GRCh38
rs770725085 Chr.16: 1361716-1361716 on Build GRCh38
rs776752687 Chr.16: 1361853-1361853 on Build GRCh38
rs200003910 Chr.16: 1361866-1361866 on Build GRCh38
rs375115440 Chr.16: 1361856-1361856 on Build GRCh38

View Full Product Details