Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35640257-35640498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD8
Entrez Gene ID
8325
Gene Name
frizzled class receptor 8
Gene Aliases
FZ-8, hFZ8
UniGene
Hs.302634
Gene Chromosome Location
Chr.10: 35638249-35641434 on Build GRCh38
Transcripts
NM_031866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4167041 c.1100_1105delCGGGCG p.A367_G368delAG Chr.10: 35640325-35640330 on Build GRCh38
6668208 c.1128C>T p.Y376Y Chr.10: 35640302-35640302 on Build GRCh38
4137974 c.954C>A p.A318A Chr.10: 35640476-35640476 on Build GRCh38
5512658 c.999G>T p.A333A Chr.10: 35640431-35640431 on Build GRCh38
1252908 c.972G>A p.S324S Chr.10: 35640458-35640458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547164495 Chr.10: 35640308-35640308 on Build GRCh38
rs762459701 Chr.10: 35640293-35640293 on Build GRCh38
rs749121600 Chr.10: 35640386-35640386 on Build GRCh38
rs771432320 Chr.10: 35640325-35640345 on Build GRCh38
rs1012527420 Chr.10: 35640343-35640348 on Build GRCh38
rs752620214 Chr.10: 35640321-35640321 on Build GRCh38
rs752279180 Chr.10: 35640478-35640478 on Build GRCh38
rs1057244884 Chr.10: 35640342-35640342 on Build GRCh38
rs998290793 Chr.10: 35640413-35640413 on Build GRCh38
rs775617182 Chr.10: 35640389-35640397 on Build GRCh38
rs747306960 Chr.10: 35640309-35640348 on Build GRCh38
rs921270517 Chr.10: 35640312-35640312 on Build GRCh38
rs762239625 Chr.10: 35640448-35640448 on Build GRCh38
rs756111900 Chr.10: 35640337-35640337 on Build GRCh38
rs754546457 Chr.10: 35640318-35640318 on Build GRCh38
rs1043326847 Chr.10: 35640346-35640346 on Build GRCh38
rs745932110 Chr.10: 35640407-35640407 on Build GRCh38
rs547056894 Chr.10: 35640338-35640338 on Build GRCh38
rs539148275 Chr.10: 35640274-35640274 on Build GRCh38
rs369427372 Chr.10: 35640322-35640322 on Build GRCh38
rs953995632 Chr.10: 35640315-35640323 on Build GRCh38
rs777191512 Chr.10: 35640328-35640342 on Build GRCh38
rs977220514 Chr.10: 35640275-35640275 on Build GRCh38
rs73270587 Chr.10: 35640422-35640422 on Build GRCh38
rs766009257 Chr.10: 35640461-35640461 on Build GRCh38
rs751000633 Chr.10: 35640317-35640317 on Build GRCh38
rs747433051 Chr.10: 35640420-35640420 on Build GRCh38
rs777084019 Chr.10: 35640290-35640290 on Build GRCh38
rs909127689 Chr.10: 35640325-35640330 on Build GRCh38
rs766029981 Chr.10: 35640302-35640302 on Build GRCh38
rs776798342 Chr.10: 35640431-35640431 on Build GRCh38
rs760114861 Chr.10: 35640340-35640345 on Build GRCh38
rs1036185969 Chr.10: 35640335-35640335 on Build GRCh38
rs759018649 Chr.10: 35640476-35640476 on Build GRCh38
rs779346880 Chr.10: 35640406-35640406 on Build GRCh38
rs772325358 Chr.10: 35640408-35640408 on Build GRCh38
rs202231423 Chr.10: 35640464-35640464 on Build GRCh38
rs757488164 Chr.10: 35640390-35640390 on Build GRCh38
rs938462784 Chr.10: 35640341-35640341 on Build GRCh38
rs536128254 Chr.10: 35640389-35640389 on Build GRCh38
rs773901621 Chr.10: 35640300-35640301 on Build GRCh38
rs765701909 Chr.10: 35640381-35640389 on Build GRCh38
rs766977488 Chr.10: 35640477-35640477 on Build GRCh38
rs566989692 Chr.10: 35640320-35640320 on Build GRCh38
rs764406352 Chr.10: 35640289-35640289 on Build GRCh38

View Full Product Details