Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46662310-46662539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920750 c.1969G>A p.A657T Chr.16: 46662341-46662341 on Build GRCh38
4569709 c.1964T>C p.L655P Chr.16: 46662346-46662346 on Build GRCh38
4185773 c.1940A>G p.E647G Chr.16: 46662370-46662370 on Build GRCh38
7151744 c.1993G>A p.D665N Chr.16: 46662317-46662317 on Build GRCh38
2920751 c.1859A>T p.D620V Chr.16: 46662451-46662451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769692699 Chr.16: 46662354-46662354 on Build GRCh38
rs780197148 Chr.16: 46662486-46662486 on Build GRCh38
rs758632241 Chr.16: 46662453-46662453 on Build GRCh38
rs138794859 Chr.16: 46662429-46662429 on Build GRCh38
rs748012697 Chr.16: 46662339-46662339 on Build GRCh38
rs1009202678 Chr.16: 46662406-46662406 on Build GRCh38
rs1038002002 Chr.16: 46662443-46662443 on Build GRCh38
rs756550058 Chr.16: 46662415-46662415 on Build GRCh38
rs569735490 Chr.16: 46662405-46662405 on Build GRCh38
rs748025800 Chr.16: 46662500-46662500 on Build GRCh38
rs145406665 Chr.16: 46662365-46662365 on Build GRCh38
rs758922774 Chr.16: 46662409-46662409 on Build GRCh38
rs201998902 Chr.16: 46662433-46662433 on Build GRCh38
rs188286943 Chr.16: 46662452-46662452 on Build GRCh38
rs757674131 Chr.16: 46662427-46662427 on Build GRCh38
rs168745 Chr.16: 46662372-46662372 on Build GRCh38
rs549129612 Chr.16: 46662487-46662487 on Build GRCh38
rs781241576 Chr.16: 46662497-46662497 on Build GRCh38
rs144997996 Chr.16: 46662398-46662398 on Build GRCh38
rs754978958 Chr.16: 46662492-46662492 on Build GRCh38
rs764695026 Chr.16: 46662416-46662416 on Build GRCh38
rs754270736 Chr.16: 46662419-46662419 on Build GRCh38

View Full Product Details