Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51049154-51049415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4381144 c.439_440delAC p.L148fs*3 Chr.18: 51049309-51049310 on Build GRCh38
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
6784668 c.428T>A p.L143H Chr.18: 51049298-51049298 on Build GRCh38
13115 c.431C>G p.S144* Chr.18: 51049301-51049301 on Build GRCh38
4275599 c.431C>G p.S144* Chr.18: 51049301-51049301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767419859 Chr.18: 51049278-51049278 on Build GRCh38
rs961003360 Chr.18: 51049223-51049223 on Build GRCh38
rs377767328 Chr.18: 51049300-51049301 on Build GRCh38
rs770463358 Chr.18: 51049370-51049370 on Build GRCh38
rs1019895180 Chr.18: 51049218-51049218 on Build GRCh38
rs758615780 Chr.18: 51049344-51049344 on Build GRCh38
rs200778699 Chr.18: 51049347-51049347 on Build GRCh38
rs762663804 Chr.18: 51049249-51049249 on Build GRCh38
rs972410674 Chr.18: 51049234-51049234 on Build GRCh38
rs377767329 Chr.18: 51049307-51049307 on Build GRCh38
rs761239224 Chr.18: 51049259-51049259 on Build GRCh38
rs779469567 Chr.18: 51049243-51049243 on Build GRCh38
rs377767327 Chr.18: 51049289-51049289 on Build GRCh38
rs201924714 Chr.18: 51049362-51049362 on Build GRCh38
rs938556033 Chr.18: 51049345-51049345 on Build GRCh38
rs864622099 Chr.18: 51049332-51049332 on Build GRCh38
rs536937954 Chr.18: 51049346-51049346 on Build GRCh38
rs773982472 Chr.18: 51049254-51049254 on Build GRCh38
rs61751987 Chr.18: 51049294-51049294 on Build GRCh38
rs763904908 Chr.18: 51049250-51049250 on Build GRCh38
rs979884322 Chr.18: 51049299-51049299 on Build GRCh38
rs750355699 Chr.18: 51049319-51049319 on Build GRCh38
rs753677989 Chr.18: 51049343-51049343 on Build GRCh38
rs193000742 Chr.18: 51049378-51049378 on Build GRCh38
rs547744892 Chr.18: 51049182-51049182 on Build GRCh38
rs746337560 Chr.18: 51049367-51049367 on Build GRCh38
rs375701524 Chr.18: 51049266-51049266 on Build GRCh38
rs766144339 Chr.18: 51049336-51049336 on Build GRCh38
rs138704821 Chr.18: 51049233-51049233 on Build GRCh38
rs757291177 Chr.18: 51049351-51049351 on Build GRCh38
rs755891338 Chr.18: 51049331-51049331 on Build GRCh38

View Full Product Details