Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100627262-100627489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4628563 c.1850C>T p.A617V Chr.7: 100627409-100627409 on Build GRCh38
3430993 c.1944C>T p.D648D Chr.7: 100627315-100627315 on Build GRCh38
7422788 c.1967A>T p.N656I Chr.7: 100627292-100627292 on Build GRCh38
3698051 c.1770C>T p.D590D Chr.7: 100627489-100627489 on Build GRCh38
1622067 c.1861_1872del12 p.A621_Q624delAVAQ Chr.7: 100627387-100627398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955254550 Chr.7: 100627458-100627458 on Build GRCh38
rs751724123 Chr.7: 100627418-100627418 on Build GRCh38
rs1004353651 Chr.7: 100627411-100627411 on Build GRCh38
rs750689567 Chr.7: 100627341-100627341 on Build GRCh38
rs753158589 Chr.7: 100627369-100627369 on Build GRCh38
rs775278479 Chr.7: 100627395-100627395 on Build GRCh38
rs544210374 Chr.7: 100627467-100627467 on Build GRCh38
rs751548369 Chr.7: 100627364-100627364 on Build GRCh38
rs80338888 Chr.7: 100627387-100627398 on Build GRCh38
rs778534889 Chr.7: 100627384-100627384 on Build GRCh38
rs766491479 Chr.7: 100627414-100627414 on Build GRCh38
rs376552157 Chr.7: 100627471-100627471 on Build GRCh38
rs370288878 Chr.7: 100627381-100627381 on Build GRCh38
rs745338834 Chr.7: 100627390-100627390 on Build GRCh38
rs376149707 Chr.7: 100627379-100627379 on Build GRCh38
rs111760099 Chr.7: 100627318-100627318 on Build GRCh38
rs759516749 Chr.7: 100627436-100627436 on Build GRCh38
rs1056668646 Chr.7: 100627397-100627397 on Build GRCh38
rs374489702 Chr.7: 100627396-100627396 on Build GRCh38
rs763294601 Chr.7: 100627413-100627413 on Build GRCh38
rs2075674 Chr.7: 100627408-100627408 on Build GRCh38
rs568798511 Chr.7: 100627302-100627302 on Build GRCh38
rs778041715 Chr.7: 100627375-100627375 on Build GRCh38
rs768578503 Chr.7: 100627406-100627406 on Build GRCh38
rs539909716 Chr.7: 100627339-100627339 on Build GRCh38
rs755561602 Chr.7: 100627365-100627365 on Build GRCh38
rs539391212 Chr.7: 100627311-100627311 on Build GRCh38
rs1021635096 Chr.7: 100627407-100627407 on Build GRCh38
rs763205379 Chr.7: 100627349-100627349 on Build GRCh38
rs772114496 Chr.7: 100627391-100627391 on Build GRCh38
rs759540608 Chr.7: 100627320-100627320 on Build GRCh38
rs770431152 Chr.7: 100627300-100627300 on Build GRCh38
rs368761907 Chr.7: 100627441-100627441 on Build GRCh38
rs1003429018 Chr.7: 100627329-100627329 on Build GRCh38
rs778131373 Chr.7: 100627451-100627451 on Build GRCh38
rs920217192 Chr.7: 100627296-100627296 on Build GRCh38
rs41295918 Chr.7: 100627357-100627357 on Build GRCh38
rs767573164 Chr.7: 100627440-100627440 on Build GRCh38
rs572997617 Chr.7: 100627325-100627325 on Build GRCh38
rs771543812 Chr.7: 100627410-100627410 on Build GRCh38
rs779001760 Chr.7: 100627350-100627350 on Build GRCh38
rs930230794 Chr.7: 100627304-100627304 on Build GRCh38
rs147286030 Chr.7: 100627447-100627447 on Build GRCh38
rs775396434 Chr.7: 100627335-100627335 on Build GRCh38
rs760383272 Chr.7: 100627336-100627336 on Build GRCh38
rs756482699 Chr.7: 100627370-100627370 on Build GRCh38

View Full Product Details