Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3589108-3589375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983073 c.4480G>A p.G1494S Chr.16: 3589158-3589158 on Build GRCh38
6144619 c.4486G>T p.G1496C Chr.16: 3589152-3589152 on Build GRCh38
6784573 c.4508C>T p.P1503L Chr.16: 3589130-3589130 on Build GRCh38
970479 c.4384G>A p.A1462T Chr.16: 3589254-3589254 on Build GRCh38
2919673 c.4407C>T p.S1469S Chr.16: 3589231-3589231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771610916 Chr.16: 3589319-3589319 on Build GRCh38
rs967369768 Chr.16: 3589314-3589314 on Build GRCh38
rs370637700 Chr.16: 3589130-3589130 on Build GRCh38
rs746008018 Chr.16: 3589143-3589143 on Build GRCh38
rs752605836 Chr.16: 3589177-3589177 on Build GRCh38
rs773941480 Chr.16: 3589261-3589261 on Build GRCh38
rs140872903 Chr.16: 3589153-3589153 on Build GRCh38
rs372321470 Chr.16: 3589211-3589211 on Build GRCh38
rs565491187 Chr.16: 3589160-3589160 on Build GRCh38
rs755887315 Chr.16: 3589178-3589178 on Build GRCh38
rs747185125 Chr.16: 3589252-3589252 on Build GRCh38
rs776332160 Chr.16: 3589337-3589337 on Build GRCh38
rs745485209 Chr.16: 3589267-3589267 on Build GRCh38
rs750635051 Chr.16: 3589248-3589248 on Build GRCh38
rs774858804 Chr.16: 3589271-3589271 on Build GRCh38
rs751653739 Chr.16: 3589247-3589257 on Build GRCh38
rs556719111 Chr.16: 3589255-3589255 on Build GRCh38
rs368388100 Chr.16: 3589294-3589294 on Build GRCh38
rs761658695 Chr.16: 3589280-3589280 on Build GRCh38
rs751536069 Chr.16: 3589295-3589295 on Build GRCh38
rs762565535 Chr.16: 3589155-3589155 on Build GRCh38
rs768653805 Chr.16: 3589276-3589276 on Build GRCh38
rs756660911 Chr.16: 3589304-3589304 on Build GRCh38
rs938457122 Chr.16: 3589223-3589223 on Build GRCh38
rs760497928 Chr.16: 3589236-3589236 on Build GRCh38
rs745883026 Chr.16: 3589202-3589202 on Build GRCh38
rs879107144 Chr.16: 3589240-3589240 on Build GRCh38
rs574214196 Chr.16: 3589235-3589235 on Build GRCh38
rs200536796 Chr.16: 3589182-3589182 on Build GRCh38
rs779970904 Chr.16: 3589251-3589251 on Build GRCh38
rs148856258 Chr.16: 3589193-3589193 on Build GRCh38
rs758409623 Chr.16: 3589203-3589203 on Build GRCh38
rs758584521 Chr.16: 3589229-3589229 on Build GRCh38
rs3810812 Chr.16: 3589138-3589138 on Build GRCh38
rs755864079 Chr.16: 3589129-3589129 on Build GRCh38
rs144037266 Chr.16: 3589257-3589257 on Build GRCh38
rs146532299 Chr.16: 3589144-3589144 on Build GRCh38
rs765485426 Chr.16: 3589245-3589245 on Build GRCh38
rs569108777 Chr.16: 3589296-3589296 on Build GRCh38
rs757128699 Chr.16: 3589244-3589244 on Build GRCh38
rs771327982 Chr.16: 3589132-3589132 on Build GRCh38
rs72778139 Chr.16: 3589229-3589229 on Build GRCh38
rs768311081 Chr.16: 3589336-3589336 on Build GRCh38
rs749082427 Chr.16: 3589260-3589260 on Build GRCh38
rs753668854 Chr.16: 3589241-3589241 on Build GRCh38
rs35603561 Chr.16: 3589312-3589312 on Build GRCh38
rs761812263 Chr.16: 3589349-3589349 on Build GRCh38
rs373300793 Chr.16: 3589291-3589291 on Build GRCh38
rs767423965 Chr.16: 3589167-3589167 on Build GRCh38
rs779332668 Chr.16: 3589198-3589198 on Build GRCh38
rs912385590 Chr.16: 3589209-3589209 on Build GRCh38
rs766416146 Chr.16: 3589234-3589244 on Build GRCh38
rs138484365 Chr.16: 3589254-3589254 on Build GRCh38
rs77718962 Chr.16: 3589300-3589300 on Build GRCh38
rs1041648763 Chr.16: 3589210-3589210 on Build GRCh38
rs769920493 Chr.16: 3589350-3589350 on Build GRCh38
rs780564269 Chr.16: 3589205-3589205 on Build GRCh38
rs139377113 Chr.16: 3589303-3589303 on Build GRCh38
rs765104618 Chr.16: 3589283-3589283 on Build GRCh38
rs769249886 Chr.16: 3589214-3589214 on Build GRCh38
rs773998146 Chr.16: 3589230-3589230 on Build GRCh38
rs1028935047 Chr.16: 3589355-3589355 on Build GRCh38
rs764189042 Chr.16: 3589237-3589237 on Build GRCh38
rs760286353 Chr.16: 3589272-3589272 on Build GRCh38
rs371090786 Chr.16: 3589299-3589299 on Build GRCh38
rs374692299 Chr.16: 3589222-3589222 on Build GRCh38
rs779769996 Chr.16: 3589325-3589325 on Build GRCh38
rs766087147 Chr.16: 3589156-3589156 on Build GRCh38
rs759045352 Chr.16: 3589232-3589232 on Build GRCh38
rs150097733 Chr.16: 3589159-3589159 on Build GRCh38
rs766956534 Chr.16: 3589234-3589234 on Build GRCh38
rs758698674 Chr.16: 3589250-3589250 on Build GRCh38
rs145444328 Chr.16: 3589189-3589189 on Build GRCh38
rs746452357 Chr.16: 3589326-3589326 on Build GRCh38
rs748571054 Chr.16: 3589226-3589226 on Build GRCh38
rs777033175 Chr.16: 3589154-3589154 on Build GRCh38
rs977339567 Chr.16: 3589324-3589324 on Build GRCh38
rs776711072 Chr.16: 3589279-3589279 on Build GRCh38

View Full Product Details