Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58548373-58548640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
UniGene
Hs.654472
Gene Chromosome Location
Chr.15: 58410754-58568952 on Build GRCh38
Transcripts
NM_000236.2 XM_005254372.1 XM_005254374.4 XM_006720502.3 XM_017022176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963342 c.1051+2T>A p.? Chr.15: 58548574-58548574 on Build GRCh38
2219419 c.982G>A p.V328I Chr.15: 58548503-58548503 on Build GRCh38
963341 c.960C>A p.R320R Chr.15: 58548481-58548481 on Build GRCh38
2219416 c.895G>A p.A299T Chr.15: 58548416-58548416 on Build GRCh38
4910465 c.944G>A p.S315N Chr.15: 58548465-58548465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776867538 Chr.15: 58548436-58548436 on Build GRCh38
rs767945380 Chr.15: 58548461-58548461 on Build GRCh38
rs142741286 Chr.15: 58548502-58548502 on Build GRCh38
rs751910773 Chr.15: 58548458-58548458 on Build GRCh38
rs751948558 Chr.15: 58548400-58548400 on Build GRCh38
rs376388170 Chr.15: 58548500-58548500 on Build GRCh38
rs780013819 Chr.15: 58548464-58548465 on Build GRCh38
rs770900492 Chr.15: 58548419-58548419 on Build GRCh38
rs764287488 Chr.15: 58548505-58548505 on Build GRCh38
rs769999239 Chr.15: 58548503-58548503 on Build GRCh38
rs553265364 Chr.15: 58548613-58548613 on Build GRCh38
rs746171308 Chr.15: 58548607-58548607 on Build GRCh38
rs760560872 Chr.15: 58548472-58548472 on Build GRCh38
rs774232279 Chr.15: 58548572-58548572 on Build GRCh38
rs145811475 Chr.15: 58548545-58548545 on Build GRCh38
rs980249517 Chr.15: 58548528-58548528 on Build GRCh38
rs753325391 Chr.15: 58548536-58548536 on Build GRCh38
rs780961693 Chr.15: 58548403-58548403 on Build GRCh38
rs995976417 Chr.15: 58548468-58548468 on Build GRCh38
rs940589138 Chr.15: 58548593-58548593 on Build GRCh38
rs773209112 Chr.15: 58548449-58548449 on Build GRCh38
rs375799764 Chr.15: 58548605-58548605 on Build GRCh38
rs756818322 Chr.15: 58548537-58548537 on Build GRCh38
rs758880495 Chr.15: 58548397-58548397 on Build GRCh38
rs747764344 Chr.15: 58548405-58548405 on Build GRCh38
rs539127704 Chr.15: 58548490-58548490 on Build GRCh38
rs763435781 Chr.15: 58548450-58548450 on Build GRCh38
rs369706830 Chr.15: 58548506-58548506 on Build GRCh38
rs773207252 Chr.15: 58548401-58548401 on Build GRCh38
rs142050387 Chr.15: 58548543-58548543 on Build GRCh38
rs768798538 Chr.15: 58548435-58548435 on Build GRCh38
rs149195239 Chr.15: 58548507-58548507 on Build GRCh38
rs984634041 Chr.15: 58548565-58548565 on Build GRCh38
rs147595392 Chr.15: 58548540-58548540 on Build GRCh38
rs559559933 Chr.15: 58548406-58548406 on Build GRCh38
rs372675212 Chr.15: 58548516-58548516 on Build GRCh38
rs766888460 Chr.15: 58548453-58548453 on Build GRCh38
rs61731970 Chr.15: 58548447-58548447 on Build GRCh38
rs765313290 Chr.15: 58548448-58548448 on Build GRCh38
rs771872430 Chr.15: 58548499-58548499 on Build GRCh38
rs758262847 Chr.15: 58548544-58548544 on Build GRCh38
rs755403316 Chr.15: 58548459-58548459 on Build GRCh38
rs774075210 Chr.15: 58548420-58548420 on Build GRCh38
rs771057642 Chr.15: 58548561-58548561 on Build GRCh38
rs200889722 Chr.15: 58548519-58548519 on Build GRCh38
rs573111255 Chr.15: 58548489-58548489 on Build GRCh38
rs372817422 Chr.15: 58548531-58548531 on Build GRCh38
rs927350688 Chr.15: 58548549-58548549 on Build GRCh38
rs528620454 Chr.15: 58548470-58548470 on Build GRCh38
rs777496353 Chr.15: 58548479-58548479 on Build GRCh38
rs372553692 Chr.15: 58548552-58548552 on Build GRCh38
rs1049029407 Chr.15: 58548417-58548417 on Build GRCh38
rs774946442 Chr.15: 58548508-58548508 on Build GRCh38
rs199867464 Chr.15: 58548424-58548424 on Build GRCh38
rs747242608 Chr.15: 58548535-58548536 on Build GRCh38
rs940558190 Chr.15: 58548434-58548434 on Build GRCh38
rs570824458 Chr.15: 58548550-58548550 on Build GRCh38
rs768828986 Chr.15: 58548615-58548616 on Build GRCh38
rs923004164 Chr.15: 58548608-58548608 on Build GRCh38
rs760190499 Chr.15: 58548602-58548602 on Build GRCh38
rs761973871 Chr.15: 58548443-58548443 on Build GRCh38
rs780285904 Chr.15: 58548398-58548398 on Build GRCh38
rs777363817 Chr.15: 58548411-58548411 on Build GRCh38
rs749244987 Chr.15: 58548415-58548415 on Build GRCh38
rs372172193 Chr.15: 58548480-58548480 on Build GRCh38
rs142388191 Chr.15: 58548518-58548518 on Build GRCh38
rs537537304 Chr.15: 58548517-58548517 on Build GRCh38

View Full Product Details