Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58548163-58548424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
UniGene
Hs.654472
Gene Chromosome Location
Chr.15: 58410754-58568952 on Build GRCh38
Transcripts
NM_000236.2 XM_005254372.1 XM_005254374.4 XM_006720502.3 XM_017022176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1373765 c.846G>A p.S282S Chr.15: 58548367-58548367 on Build GRCh38
6347464 c.809-7G>T p.? Chr.15: 58548323-58548323 on Build GRCh38
4055892 c.837C>T p.H279H Chr.15: 58548358-58548358 on Build GRCh38
2219416 c.895G>A p.A299T Chr.15: 58548416-58548416 on Build GRCh38
1517380 c.841C>T p.R281* Chr.15: 58548362-58548362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74018855 Chr.15: 58548283-58548283 on Build GRCh38
rs757817039 Chr.15: 58548315-58548315 on Build GRCh38
rs1018732469 Chr.15: 58548217-58548217 on Build GRCh38
rs761575901 Chr.15: 58548383-58548383 on Build GRCh38
rs559559933 Chr.15: 58548406-58548406 on Build GRCh38
rs779366465 Chr.15: 58548316-58548316 on Build GRCh38
rs758719680 Chr.15: 58548327-58548327 on Build GRCh38
rs917225755 Chr.15: 58548359-58548359 on Build GRCh38
rs979078435 Chr.15: 58548311-58548311 on Build GRCh38
rs1007543 Chr.15: 58548251-58548251 on Build GRCh38
rs751948558 Chr.15: 58548400-58548400 on Build GRCh38
rs774131745 Chr.15: 58548368-58548368 on Build GRCh38
rs546100913 Chr.15: 58548192-58548192 on Build GRCh38
rs1004531528 Chr.15: 58548197-58548197 on Build GRCh38
rs753913266 Chr.15: 58548307-58548307 on Build GRCh38
rs762938418 Chr.15: 58548367-58548367 on Build GRCh38
rs553254820 Chr.15: 58548250-58548250 on Build GRCh38
rs139784245 Chr.15: 58548373-58548373 on Build GRCh38
rs541958282 Chr.15: 58548263-58548263 on Build GRCh38
rs901604829 Chr.15: 58548241-58548241 on Build GRCh38
rs371853675 Chr.15: 58548337-58548337 on Build GRCh38
rs750823091 Chr.15: 58548325-58548325 on Build GRCh38
rs780961693 Chr.15: 58548403-58548403 on Build GRCh38
rs200613217 Chr.15: 58548363-58548363 on Build GRCh38
rs780626906 Chr.15: 58548342-58548342 on Build GRCh38
rs780417026 Chr.15: 58548331-58548331 on Build GRCh38
rs146299102 Chr.15: 58548358-58548358 on Build GRCh38
rs121912502 Chr.15: 58548387-58548387 on Build GRCh38
rs758880495 Chr.15: 58548397-58548397 on Build GRCh38
rs747764344 Chr.15: 58548405-58548405 on Build GRCh38
rs747641881 Chr.15: 58548349-58548349 on Build GRCh38
rs760898380 Chr.15: 58548291-58548291 on Build GRCh38
rs371200300 Chr.15: 58548191-58548191 on Build GRCh38
rs927182594 Chr.15: 58548392-58548392 on Build GRCh38
rs773207252 Chr.15: 58548401-58548401 on Build GRCh38
rs780285904 Chr.15: 58548398-58548398 on Build GRCh38
rs148453176 Chr.15: 58548336-58548336 on Build GRCh38
rs765258214 Chr.15: 58548388-58548388 on Build GRCh38
rs369378374 Chr.15: 58548382-58548382 on Build GRCh38
rs764088950 Chr.15: 58548298-58548298 on Build GRCh38

View Full Product Details