Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37850247-37850508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1089437 c.91+1G>A p.? Chr.7: 37850436-37850436 on Build GRCh38
1450586 c.9_10insA p.R6fs*11 Chr.7: 37850275-37850276 on Build GRCh38
6028283 c.10_11insA p.R6fs*11 Chr.7: 37850276-37850277 on Build GRCh38
3880846 c.18A>G p.R6R Chr.7: 37850284-37850284 on Build GRCh38
3638511 c.17G>A p.R6Q Chr.7: 37850283-37850283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750045501 Chr.7: 37850282-37850282 on Build GRCh38
rs555707678 Chr.7: 37850336-37850336 on Build GRCh38
rs771877216 Chr.7: 37850448-37850448 on Build GRCh38
rs750902815 Chr.7: 37850378-37850378 on Build GRCh38
rs373353542 Chr.7: 37850291-37850291 on Build GRCh38
rs750630112 Chr.7: 37850417-37850417 on Build GRCh38
rs752092011 Chr.7: 37850401-37850401 on Build GRCh38
rs765880975 Chr.7: 37850369-37850369 on Build GRCh38
rs772021065 Chr.7: 37850357-37850357 on Build GRCh38
rs757623095 Chr.7: 37850300-37850300 on Build GRCh38
rs374519602 Chr.7: 37850470-37850470 on Build GRCh38
rs778106704 Chr.7: 37850296-37850296 on Build GRCh38
rs924858337 Chr.7: 37850391-37850391 on Build GRCh38
rs141405304 Chr.7: 37850450-37850450 on Build GRCh38
rs748116097 Chr.7: 37850445-37850445 on Build GRCh38
rs201220646 Chr.7: 37850388-37850388 on Build GRCh38
rs369098657 Chr.7: 37850297-37850297 on Build GRCh38
rs764751735 Chr.7: 37850475-37850475 on Build GRCh38
rs1050366765 Chr.7: 37850447-37850447 on Build GRCh38
rs757570040 Chr.7: 37850404-37850404 on Build GRCh38
rs774819055 Chr.7: 37850476-37850476 on Build GRCh38
rs760293308 Chr.7: 37850368-37850368 on Build GRCh38
rs772869964 Chr.7: 37850364-37850364 on Build GRCh38
rs770546236 Chr.7: 37850453-37850453 on Build GRCh38
rs763436203 Chr.7: 37850386-37850386 on Build GRCh38
rs753590172 Chr.7: 37850296-37850296 on Build GRCh38
rs372678302 Chr.7: 37850318-37850318 on Build GRCh38
rs776071598 Chr.7: 37850380-37850380 on Build GRCh38
rs188238656 Chr.7: 37850311-37850311 on Build GRCh38
rs372718997 Chr.7: 37850486-37850486 on Build GRCh38
rs377033672 Chr.7: 37850320-37850320 on Build GRCh38
rs762350734 Chr.7: 37850477-37850477 on Build GRCh38
rs780172047 Chr.7: 37850426-37850426 on Build GRCh38
rs765828426 Chr.7: 37850290-37850290 on Build GRCh38
rs756360013 Chr.7: 37850418-37850418 on Build GRCh38
rs199920317 Chr.7: 37850432-37850432 on Build GRCh38
rs764519450 Chr.7: 37850395-37850395 on Build GRCh38
rs984892337 Chr.7: 37850340-37850340 on Build GRCh38
rs746669564 Chr.7: 37850451-37850451 on Build GRCh38
rs2722371 Chr.7: 37850479-37850479 on Build GRCh38
rs746181533 Chr.7: 37850314-37850314 on Build GRCh38
rs376117068 Chr.7: 37850428-37850428 on Build GRCh38
rs981731927 Chr.7: 37850278-37850278 on Build GRCh38
rs767922928 Chr.7: 37850478-37850478 on Build GRCh38
rs374040658 Chr.7: 37850408-37850408 on Build GRCh38
rs768774717 Chr.7: 37850327-37850327 on Build GRCh38
rs747880885 Chr.7: 37850352-37850352 on Build GRCh38
rs778952400 Chr.7: 37850434-37850434 on Build GRCh38
rs759219304 Chr.7: 37850420-37850430 on Build GRCh38
rs934943362 Chr.7: 37850397-37850397 on Build GRCh38
rs199576209 Chr.7: 37850283-37850283 on Build GRCh38
rs776350841 Chr.7: 37850461-37850461 on Build GRCh38

View Full Product Details