Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144512481-144512706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1623786 c.2855G>C p.G952A Chr.8: 144512672-144512672 on Build GRCh38
3834301 c.2841G>C p.L947L Chr.8: 144512686-144512686 on Build GRCh38
5759838 c.2894C>A p.P965H Chr.8: 144512553-144512553 on Build GRCh38
117757 c.2856C>G p.G952G Chr.8: 144512671-144512671 on Build GRCh38
383585 c.2924C>T p.P975L Chr.8: 144512523-144512523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200998618 Chr.8: 144512573-144512573 on Build GRCh38
rs772780752 Chr.8: 144512540-144512540 on Build GRCh38
rs577462627 Chr.8: 144512642-144512642 on Build GRCh38
rs746107365 Chr.8: 144512514-144512514 on Build GRCh38
rs370549048 Chr.8: 144512681-144512681 on Build GRCh38
rs368081792 Chr.8: 144512658-144512658 on Build GRCh38
rs755461118 Chr.8: 144512588-144512588 on Build GRCh38
rs752697206 Chr.8: 144512567-144512567 on Build GRCh38
rs775475361 Chr.8: 144512679-144512679 on Build GRCh38
rs746619624 Chr.8: 144512611-144512611 on Build GRCh38
rs544576633 Chr.8: 144512579-144512579 on Build GRCh38
rs33972310 Chr.8: 144512557-144512557 on Build GRCh38
rs771581851 Chr.8: 144512537-144512537 on Build GRCh38
rs780253117 Chr.8: 144512581-144512581 on Build GRCh38
rs772800634 Chr.8: 144512601-144512601 on Build GRCh38
rs587778645 Chr.8: 144512512-144512512 on Build GRCh38
rs770655093 Chr.8: 144512614-144512614 on Build GRCh38
rs760402848 Chr.8: 144512541-144512541 on Build GRCh38
rs376955025 Chr.8: 144512615-144512615 on Build GRCh38
rs776993029 Chr.8: 144512505-144512506 on Build GRCh38
rs757541591 Chr.8: 144512506-144512506 on Build GRCh38
rs770474816 Chr.8: 144512669-144512669 on Build GRCh38
rs770014424 Chr.8: 144512515-144512515 on Build GRCh38
rs759359138 Chr.8: 144512559-144512559 on Build GRCh38
rs763705281 Chr.8: 144512614-144512615 on Build GRCh38
rs766038992 Chr.8: 144512553-144512553 on Build GRCh38
rs762544755 Chr.8: 144512603-144512614 on Build GRCh38
rs973332338 Chr.8: 144512602-144512602 on Build GRCh38
rs750560547 Chr.8: 144512637-144512637 on Build GRCh38
rs769664095 Chr.8: 144512678-144512678 on Build GRCh38
rs545478910 Chr.8: 144512676-144512676 on Build GRCh38
rs779276650 Chr.8: 144512650-144512650 on Build GRCh38
rs759319751 Chr.8: 144512623-144512623 on Build GRCh38
rs556430854 Chr.8: 144512589-144512589 on Build GRCh38
rs780492921 Chr.8: 144512518-144512518 on Build GRCh38
rs746594220 Chr.8: 144512666-144512666 on Build GRCh38
rs35070885 Chr.8: 144512521-144512521 on Build GRCh38
rs775156254 Chr.8: 144512599-144512600 on Build GRCh38
rs776281689 Chr.8: 144512672-144512672 on Build GRCh38
rs765131003 Chr.8: 144512627-144512627 on Build GRCh38
rs766684128 Chr.8: 144512647-144512647 on Build GRCh38
rs771526891 Chr.8: 144512599-144512599 on Build GRCh38
rs373651108 Chr.8: 144512648-144512648 on Build GRCh38
rs761367235 Chr.8: 144512534-144512534 on Build GRCh38
rs756288212 Chr.8: 144512639-144512639 on Build GRCh38
rs555930479 Chr.8: 144512622-144512622 on Build GRCh38
rs751422368 Chr.8: 144512657-144512658 on Build GRCh38
rs748601844 Chr.8: 144512653-144512653 on Build GRCh38
rs764586373 Chr.8: 144512591-144512592 on Build GRCh38
rs559089481 Chr.8: 144512582-144512582 on Build GRCh38
rs773849437 Chr.8: 144512525-144512525 on Build GRCh38
rs769194048 Chr.8: 144512523-144512523 on Build GRCh38
rs775528539 Chr.8: 144512630-144512630 on Build GRCh38
rs578139954 Chr.8: 144512633-144512633 on Build GRCh38
rs757341042 Chr.8: 144512568-144512568 on Build GRCh38
rs776537316 Chr.8: 144512555-144512555 on Build GRCh38
rs1015300741 Chr.8: 144512572-144512572 on Build GRCh38
rs548485975 Chr.8: 144512656-144512656 on Build GRCh38
rs748654853 Chr.8: 144512595-144512595 on Build GRCh38
rs1004279093 Chr.8: 144512566-144512566 on Build GRCh38
rs759100987 Chr.8: 144512584-144512585 on Build GRCh38
rs750563407 Chr.8: 144512577-144512577 on Build GRCh38
rs765331236 Chr.8: 144512564-144512564 on Build GRCh38
rs370547126 Chr.8: 144512634-144512634 on Build GRCh38
rs758952928 Chr.8: 144512654-144512654 on Build GRCh38
rs1022684920 Chr.8: 144512598-144512598 on Build GRCh38
rs560394637 Chr.8: 144512680-144512680 on Build GRCh38
rs368710171 Chr.8: 144512557-144512557 on Build GRCh38

View Full Product Details