Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3589666-3589927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2919685 c.3950C>T p.P1317L Chr.16: 3589688-3589688 on Build GRCh38
4129051 c.3942G>A p.Q1314Q Chr.16: 3589696-3589696 on Build GRCh38
4129050 c.3943A>G p.T1315A Chr.16: 3589695-3589695 on Build GRCh38
1377773 c.3721C>T p.R1241C Chr.16: 3589917-3589917 on Build GRCh38
6784575 c.3841G>A p.G1281R Chr.16: 3589797-3589797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149011965 Chr.16: 3589781-3589781 on Build GRCh38
rs556682517 Chr.16: 3589712-3589712 on Build GRCh38
rs3810814 Chr.16: 3589876-3589876 on Build GRCh38
rs749433774 Chr.16: 3589779-3589779 on Build GRCh38
rs749975141 Chr.16: 3589758-3589758 on Build GRCh38
rs759828401 Chr.16: 3589731-3589731 on Build GRCh38
rs376276225 Chr.16: 3589807-3589807 on Build GRCh38
rs202148391 Chr.16: 3589880-3589880 on Build GRCh38
rs376232540 Chr.16: 3589900-3589900 on Build GRCh38
rs146837290 Chr.16: 3589727-3589727 on Build GRCh38
rs374311917 Chr.16: 3589705-3589705 on Build GRCh38
rs755975486 Chr.16: 3589866-3589866 on Build GRCh38
rs773956906 Chr.16: 3589860-3589860 on Build GRCh38
rs779426291 Chr.16: 3589832-3589832 on Build GRCh38
rs142040192 Chr.16: 3589698-3589698 on Build GRCh38
rs140103960 Chr.16: 3589790-3589790 on Build GRCh38
rs763914156 Chr.16: 3589742-3589743 on Build GRCh38
rs754996241 Chr.16: 3589797-3589797 on Build GRCh38
rs375172024 Chr.16: 3589688-3589688 on Build GRCh38
rs370663234 Chr.16: 3589863-3589863 on Build GRCh38
rs201294529 Chr.16: 3589895-3589895 on Build GRCh38
rs770020797 Chr.16: 3589710-3589710 on Build GRCh38
rs760555181 Chr.16: 3589724-3589725 on Build GRCh38
rs754761366 Chr.16: 3589766-3589766 on Build GRCh38
rs1040333386 Chr.16: 3589792-3589792 on Build GRCh38
rs936127000 Chr.16: 3589767-3589767 on Build GRCh38
rs775711964 Chr.16: 3589838-3589838 on Build GRCh38
rs769032315 Chr.16: 3589897-3589897 on Build GRCh38
rs779981907 Chr.16: 3589763-3589763 on Build GRCh38
rs563781078 Chr.16: 3589828-3589828 on Build GRCh38
rs766299864 Chr.16: 3589796-3589796 on Build GRCh38
rs374481298 Chr.16: 3589752-3589752 on Build GRCh38
rs756446820 Chr.16: 3589777-3589777 on Build GRCh38
rs767189475 Chr.16: 3589798-3589798 on Build GRCh38
rs762456970 Chr.16: 3589795-3589795 on Build GRCh38
rs768867319 Chr.16: 3589701-3589701 on Build GRCh38
rs775548839 Chr.16: 3589717-3589719 on Build GRCh38
rs748912324 Chr.16: 3589904-3589904 on Build GRCh38
rs772673695 Chr.16: 3589837-3589837 on Build GRCh38
rs780617305 Chr.16: 3589884-3589884 on Build GRCh38
rs769371909 Chr.16: 3589847-3589847 on Build GRCh38
rs147600630 Chr.16: 3589687-3589687 on Build GRCh38
rs763731687 Chr.16: 3589732-3589732 on Build GRCh38
rs757548615 Chr.16: 3589870-3589870 on Build GRCh38
rs201422743 Chr.16: 3589839-3589839 on Build GRCh38
rs77699867 Chr.16: 3589855-3589855 on Build GRCh38
rs758320008 Chr.16: 3589761-3589761 on Build GRCh38
rs367741978 Chr.16: 3589831-3589831 on Build GRCh38
rs1025512712 Chr.16: 3589835-3589835 on Build GRCh38
rs377243383 Chr.16: 3589745-3589745 on Build GRCh38
rs373107728 Chr.16: 3589788-3589788 on Build GRCh38
rs574928535 Chr.16: 3589791-3589791 on Build GRCh38
rs369714443 Chr.16: 3589699-3589699 on Build GRCh38
rs372991604 Chr.16: 3589892-3589892 on Build GRCh38
rs753464234 Chr.16: 3589735-3589735 on Build GRCh38
rs112596894 Chr.16: 3589770-3589770 on Build GRCh38
rs756389198 Chr.16: 3589811-3589811 on Build GRCh38
rs750580998 Chr.16: 3589877-3589877 on Build GRCh38
rs146054214 Chr.16: 3589864-3589864 on Build GRCh38
rs3810813 Chr.16: 3589826-3589826 on Build GRCh38
rs763524714 Chr.16: 3589725-3589725 on Build GRCh38
rs772652088 Chr.16: 3589794-3589794 on Build GRCh38
rs374056556 Chr.16: 3589856-3589856 on Build GRCh38
rs771503932 Chr.16: 3589780-3589780 on Build GRCh38
rs916888522 Chr.16: 3589865-3589865 on Build GRCh38
rs199958007 Chr.16: 3589836-3589836 on Build GRCh38
rs200928592 Chr.16: 3589789-3589789 on Build GRCh38
rs141860134 Chr.16: 3589857-3589857 on Build GRCh38
rs751302297 Chr.16: 3589765-3589765 on Build GRCh38
rs188808130 Chr.16: 3589757-3589757 on Build GRCh38
rs991818380 Chr.16: 3589854-3589854 on Build GRCh38
rs140254478 Chr.16: 3589726-3589726 on Build GRCh38
rs201037631 Chr.16: 3589868-3589868 on Build GRCh38
rs776294939 Chr.16: 3589893-3589893 on Build GRCh38
rs200008765 Chr.16: 3589704-3589704 on Build GRCh38
rs978074181 Chr.16: 3589899-3589899 on Build GRCh38

View Full Product Details