Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16986698-16986923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4785781 c.3003C>T p.Y1001Y Chr.1: 16986905-16986905 on Build GRCh38
1687074 c.3217C>T p.R1073W Chr.1: 16986823-16986823 on Build GRCh38
4025161 c.3233A>G p.N1078S Chr.1: 16986807-16986807 on Build GRCh38
5783752 c.3214C>T p.R1072C Chr.1: 16986826-16986826 on Build GRCh38
1295497 c.3138G>C p.E1046D Chr.1: 16986902-16986902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971333396 Chr.1: 16986823-16986823 on Build GRCh38
rs137877665 Chr.1: 16986833-16986833 on Build GRCh38
rs769818426 Chr.1: 16986774-16986774 on Build GRCh38
rs369278996 Chr.1: 16986792-16986792 on Build GRCh38
rs779674096 Chr.1: 16986757-16986757 on Build GRCh38
rs974089234 Chr.1: 16986742-16986742 on Build GRCh38
rs758287038 Chr.1: 16986749-16986750 on Build GRCh38
rs137853967 Chr.1: 16986864-16986864 on Build GRCh38
rs373315947 Chr.1: 16986789-16986789 on Build GRCh38
rs377431904 Chr.1: 16986847-16986847 on Build GRCh38
rs116456892 Chr.1: 16986784-16986784 on Build GRCh38
rs376475762 Chr.1: 16986785-16986785 on Build GRCh38
rs372993383 Chr.1: 16986818-16986818 on Build GRCh38
rs1046254851 Chr.1: 16986876-16986876 on Build GRCh38
rs9435659 Chr.1: 16986848-16986848 on Build GRCh38
rs777755640 Chr.1: 16986826-16986826 on Build GRCh38
rs1047701202 Chr.1: 16986764-16986764 on Build GRCh38
rs371220869 Chr.1: 16986825-16986825 on Build GRCh38
rs944188699 Chr.1: 16986883-16986883 on Build GRCh38
rs937351387 Chr.1: 16986734-16986734 on Build GRCh38
rs747907490 Chr.1: 16986822-16986822 on Build GRCh38
rs567315420 Chr.1: 16986795-16986795 on Build GRCh38
rs199661502 Chr.1: 16986753-16986760 on Build GRCh38
rs377380258 Chr.1: 16986753-16986764 on Build GRCh38
rs757851198 Chr.1: 16986794-16986794 on Build GRCh38
rs762212020 Chr.1: 16986869-16986869 on Build GRCh38
rs56146840 Chr.1: 16986788-16986788 on Build GRCh38
rs140394047 Chr.1: 16986881-16986881 on Build GRCh38
rs760935912 Chr.1: 16986863-16986863 on Build GRCh38
rs369142791 Chr.1: 16986749-16986749 on Build GRCh38
rs750963832 Chr.1: 16986895-16986895 on Build GRCh38
rs768397313 Chr.1: 16986772-16986772 on Build GRCh38
rs927333814 Chr.1: 16986726-16986726 on Build GRCh38
rs9435735 Chr.1: 16986719-16986719 on Build GRCh38
rs930131952 Chr.1: 16986762-16986762 on Build GRCh38
rs369718815 Chr.1: 16986773-16986773 on Build GRCh38
rs763038059 Chr.1: 16986781-16986781 on Build GRCh38
rs868043973 Chr.1: 16986812-16986812 on Build GRCh38
rs371155708 Chr.1: 16986857-16986857 on Build GRCh38
rs747617559 Chr.1: 16986886-16986887 on Build GRCh38
rs56126202 Chr.1: 16986896-16986896 on Build GRCh38
rs746638178 Chr.1: 16986771-16986771 on Build GRCh38
rs1020189579 Chr.1: 16986717-16986717 on Build GRCh38
rs764988645 Chr.1: 16986834-16986834 on Build GRCh38
rs761031832 Chr.1: 16986790-16986790 on Build GRCh38
rs975127788 Chr.1: 16986715-16986715 on Build GRCh38
rs143483351 Chr.1: 16986750-16986761 on Build GRCh38
rs764410859 Chr.1: 16986865-16986865 on Build GRCh38
rs777045615 Chr.1: 16986866-16986866 on Build GRCh38
rs376397035 Chr.1: 16986791-16986791 on Build GRCh38
rs951584451 Chr.1: 16986851-16986851 on Build GRCh38
rs375233425 Chr.1: 16986725-16986725 on Build GRCh38
rs773291692 Chr.1: 16986776-16986776 on Build GRCh38
rs976061203 Chr.1: 16986793-16986793 on Build GRCh38
rs528997540 Chr.1: 16986718-16986718 on Build GRCh38
rs780992643 Chr.1: 16986819-16986819 on Build GRCh38
rs774238872 Chr.1: 16986835-16986835 on Build GRCh38
rs1057519290 Chr.1: 16986886-16986888 on Build GRCh38

View Full Product Details