Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1362047-1362284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672465 c.485C>T p.T162M Chr.16: 1362279-1362279 on Build GRCh38
5470440 c.486G>A p.T162T Chr.16: 1362280-1362280 on Build GRCh38
1253377 c.425G>A p.C142Y Chr.16: 1362219-1362219 on Build GRCh38
1208435 c.340G>A p.A114T Chr.16: 1362060-1362060 on Build GRCh38
1301638 c.405G>A p.Q135Q Chr.16: 1362125-1362125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767760180 Chr.16: 1362183-1362183 on Build GRCh38
rs200975355 Chr.16: 1362067-1362067 on Build GRCh38
rs747291812 Chr.16: 1362082-1362082 on Build GRCh38
rs767633692 Chr.16: 1362247-1362247 on Build GRCh38
rs752836392 Chr.16: 1362070-1362070 on Build GRCh38
rs763065729 Chr.16: 1362251-1362251 on Build GRCh38
rs528538033 Chr.16: 1362192-1362192 on Build GRCh38
rs281864956 Chr.16: 1362239-1362239 on Build GRCh38
rs773268405 Chr.16: 1362249-1362249 on Build GRCh38
rs760265920 Chr.16: 1362172-1362173 on Build GRCh38
rs562132501 Chr.16: 1362242-1362242 on Build GRCh38
rs564859061 Chr.16: 1362265-1362265 on Build GRCh38
rs756379978 Chr.16: 1362216-1362216 on Build GRCh38
rs772949824 Chr.16: 1362170-1362170 on Build GRCh38
rs374890181 Chr.16: 1362234-1362234 on Build GRCh38
rs753205380 Chr.16: 1362120-1362120 on Build GRCh38
rs200741370 Chr.16: 1362114-1362114 on Build GRCh38
rs753100529 Chr.16: 1362159-1362160 on Build GRCh38
rs758452962 Chr.16: 1362077-1362077 on Build GRCh38
rs200508759 Chr.16: 1362176-1362176 on Build GRCh38
rs778657195 Chr.16: 1362198-1362198 on Build GRCh38
rs751676968 Chr.16: 1362178-1362178 on Build GRCh38
rs760954235 Chr.16: 1362250-1362250 on Build GRCh38
rs757509701 Chr.16: 1362144-1362144 on Build GRCh38
rs768465491 Chr.16: 1362153-1362153 on Build GRCh38
rs750858322 Chr.16: 1362172-1362172 on Build GRCh38
rs765865746 Chr.16: 1362118-1362118 on Build GRCh38
rs775359476 Chr.16: 1362096-1362096 on Build GRCh38
rs193302849 Chr.16: 1362067-1362069 on Build GRCh38
rs753679575 Chr.16: 1362254-1362254 on Build GRCh38
rs768086598 Chr.16: 1362133-1362134 on Build GRCh38
rs761835152 Chr.16: 1362179-1362179 on Build GRCh38
rs372319167 Chr.16: 1362102-1362102 on Build GRCh38
rs141870165 Chr.16: 1362095-1362095 on Build GRCh38
rs530486639 Chr.16: 1362217-1362217 on Build GRCh38
rs781036850 Chr.16: 1362092-1362092 on Build GRCh38
rs1033139347 Chr.16: 1362124-1362124 on Build GRCh38
rs138018487 Chr.16: 1362266-1362266 on Build GRCh38
rs940572438 Chr.16: 1362221-1362221 on Build GRCh38
rs368718738 Chr.16: 1362193-1362193 on Build GRCh38
rs777887031 Chr.16: 1362081-1362081 on Build GRCh38
rs747713846 Chr.16: 1362200-1362200 on Build GRCh38
rs775985395 Chr.16: 1362233-1362233 on Build GRCh38
rs751575030 Chr.16: 1362135-1362135 on Build GRCh38
rs747545433 Chr.16: 1362161-1362161 on Build GRCh38
rs749298719 Chr.16: 1362151-1362151 on Build GRCh38
rs1005797412 Chr.16: 1362185-1362185 on Build GRCh38
rs375386449 Chr.16: 1362146-1362146 on Build GRCh38
rs781337088 Chr.16: 1362145-1362145 on Build GRCh38
rs780619430 Chr.16: 1362256-1362256 on Build GRCh38
rs906630870 Chr.16: 1362160-1362160 on Build GRCh38
rs774898087 Chr.16: 1362167-1362167 on Build GRCh38
rs147109185 Chr.16: 1362244-1362244 on Build GRCh38
rs761584357 Chr.16: 1362108-1362108 on Build GRCh38
rs576804198 Chr.16: 1362154-1362154 on Build GRCh38
rs73487805 Chr.16: 1362189-1362189 on Build GRCh38
rs763335225 Chr.16: 1362125-1362125 on Build GRCh38
rs11554983 Chr.16: 1362115-1362115 on Build GRCh38
rs763669202 Chr.16: 1362176-1362183 on Build GRCh38
rs760008789 Chr.16: 1362171-1362171 on Build GRCh38
rs193302850 Chr.16: 1362099-1362111 on Build GRCh38
rs138297609 Chr.16: 1362064-1362064 on Build GRCh38
rs748782055 Chr.16: 1362101-1362101 on Build GRCh38
rs756307321 Chr.16: 1362190-1362190 on Build GRCh38
rs757394606 Chr.16: 1362086-1362086 on Build GRCh38
rs771863638 Chr.16: 1362163-1362163 on Build GRCh38
rs769233183 Chr.16: 1362235-1362235 on Build GRCh38
rs779795759 Chr.16: 1362149-1362149 on Build GRCh38
rs554418793 Chr.16: 1362109-1362109 on Build GRCh38
rs1000273462 Chr.16: 1362262-1362262 on Build GRCh38
rs113388753 Chr.16: 1362258-1362258 on Build GRCh38
rs775693232 Chr.16: 1362175-1362175 on Build GRCh38
rs754679713 Chr.16: 1362255-1362255 on Build GRCh38
rs764279342 Chr.16: 1362128-1362128 on Build GRCh38

View Full Product Details