Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161973197-161973470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6737755 c.619-1G>T p.? Chr.6: 161973418-161973418 on Build GRCh38
3393967 c.729C>T p.D243D Chr.6: 161973307-161973307 on Build GRCh38
741235 c.685A>T p.I229F Chr.6: 161973351-161973351 on Build GRCh38
3860255 c.709A>G p.T237A Chr.6: 161973327-161973327 on Build GRCh38
4711141 c.700C>T p.R234W Chr.6: 161973336-161973336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776313115 Chr.6: 161973447-161973447 on Build GRCh38
rs751272531 Chr.6: 161973367-161973367 on Build GRCh38
rs777608834 Chr.6: 161973287-161973287 on Build GRCh38
rs758048810 Chr.6: 161973349-161973349 on Build GRCh38
rs759422627 Chr.6: 161973340-161973340 on Build GRCh38
rs754452026 Chr.6: 161973267-161973267 on Build GRCh38
rs137853060 Chr.6: 161973403-161973403 on Build GRCh38
rs760135330 Chr.6: 161973326-161973326 on Build GRCh38
rs761944496 Chr.6: 161973437-161973437 on Build GRCh38
rs781443859 Chr.6: 161973383-161973383 on Build GRCh38
rs757579280 Chr.6: 161973377-161973377 on Build GRCh38
rs539815495 Chr.6: 161973365-161973365 on Build GRCh38
rs201881954 Chr.6: 161973264-161973264 on Build GRCh38
rs983130934 Chr.6: 161973343-161973343 on Build GRCh38
rs764754559 Chr.6: 161973344-161973344 on Build GRCh38
rs770718178 Chr.6: 161973443-161973443 on Build GRCh38
rs769882260 Chr.6: 161973316-161973316 on Build GRCh38
rs146173584 Chr.6: 161973309-161973309 on Build GRCh38
rs528962770 Chr.6: 161973231-161973231 on Build GRCh38
rs565463567 Chr.6: 161973268-161973268 on Build GRCh38
rs571490973 Chr.6: 161973347-161973347 on Build GRCh38
rs768832012 Chr.6: 161973320-161973320 on Build GRCh38
rs137853054 Chr.6: 161973317-161973317 on Build GRCh38
rs987539311 Chr.6: 161973293-161973293 on Build GRCh38
rs368076010 Chr.6: 161973236-161973236 on Build GRCh38
rs144032774 Chr.6: 161973335-161973335 on Build GRCh38
rs770182456 Chr.6: 161973397-161973397 on Build GRCh38
rs774496098 Chr.6: 161973429-161973429 on Build GRCh38
rs551801791 Chr.6: 161973373-161973373 on Build GRCh38
rs137853058 Chr.6: 161973401-161973401 on Build GRCh38
rs759416147 Chr.6: 161973448-161973448 on Build GRCh38
rs989568138 Chr.6: 161973382-161973387 on Build GRCh38
rs373822092 Chr.6: 161973336-161973336 on Build GRCh38
rs886061238 Chr.6: 161973330-161973330 on Build GRCh38
rs766332636 Chr.6: 161973263-161973263 on Build GRCh38
rs114974496 Chr.6: 161973322-161973322 on Build GRCh38
rs768779161 Chr.6: 161973417-161973417 on Build GRCh38
rs938983242 Chr.6: 161973413-161973413 on Build GRCh38
rs548730727 Chr.6: 161973247-161973247 on Build GRCh38
rs750192653 Chr.6: 161973254-161973254 on Build GRCh38
rs755939170 Chr.6: 161973258-161973258 on Build GRCh38
rs758101954 Chr.6: 161973271-161973271 on Build GRCh38
rs1023253858 Chr.6: 161973232-161973232 on Build GRCh38
rs747762203 Chr.6: 161973269-161973269 on Build GRCh38
rs747376218 Chr.6: 161973304-161973304 on Build GRCh38
rs202212928 Chr.6: 161973363-161973363 on Build GRCh38
rs993941105 Chr.6: 161973272-161973272 on Build GRCh38
rs894329504 Chr.6: 161973255-161973255 on Build GRCh38
rs969335435 Chr.6: 161973233-161973233 on Build GRCh38
rs746368282 Chr.6: 161973311-161973311 on Build GRCh38
rs771259513 Chr.6: 161973306-161973306 on Build GRCh38
rs956142276 Chr.6: 161973282-161973282 on Build GRCh38
rs746340817 Chr.6: 161973391-161973391 on Build GRCh38
rs928405958 Chr.6: 161973228-161973228 on Build GRCh38
rs777129050 Chr.6: 161973292-161973292 on Build GRCh38

View Full Product Details