Gene Transcripts

Gene Symbol
Wdr92
Entrez Gene ID
103784
Gene Name
WD repeat domain 92
Gene Aliases
AI553587, HZGJ
Chromosome Location
Chr. 11: 17211893 - 17235200 on Build GRCm38
UniGene ID
Mm.298132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178909.4 NP_849240.1 2 305
GenBank mRNA AK033294.1 - 2 251
AK047691.1 - 2 280
AK088218.1 - 2 280
AK142228.1 - 2 251
BC096540.1 - 2 234 6403681
BY794211.1 - 2 306

View Full Product Details