Gene Transcripts

Gene Symbol
Epdr1
Entrez Gene ID
105298
Gene Name
ependymin related protein 1 (zebrafish)
Gene Aliases
AU040950, AV006210, Epdm2, Epdr2, MERP-1, MERP-2, MERP2, Ucc1
Chromosome Location
Chr. 13: 19591708 - 19619952 on Build GRCm38
UniGene ID
Mm.275054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134065.4 NP_598826.3 2 661
GenBank mRNA AF353717.1 - 2 474
AK034083.1 - 2 630
AK038733.1 - 2 636
AK046185.1 - 2 636
AK160413.1 - 2 596
AK166252.1 - 2 602
BC014708.1 - 2 616 4223134

View Full Product Details