Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014976.2 | NP_001014976.1 | 27 | 5899 | |
GenBank mRNA | AK129072.1 | - | 27 | 5866 | |
AY588413.1 | - | 27 | 5837 | ||
BC082603.1 | - | 27 | 5899 | 30668743 | |
BC145844.1 | - | 27 | 5870 | 40131147 | |
BC145846.1 | - | 27 | 5816 | 40131149 |