Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
110172
Gene Name
solute carrier family 35, member B1
Gene Aliases
Ugalt2
Chromosome Location
Chr. 11: 95384891 - 95391776 on Build GRCm38
UniGene ID
Mm.4593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016752.1 NP_058032.1 5 569
GenBank mRNA AK002961.1 - 5 563
AK007711.1 - 5 575
AK160124.1 - 5 635
D87990.1 - 5 569

View Full Product Details