Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110794.1 | NP_001104264.1 | 11 | 1376 | |
NM_009674.3 | NP_033804.2 | 11 | 1491 | ||
GenBank mRNA | AK032013.1 | - | 11 | 1267 | |
AK077579.1 | - | 11 | 1184 | ||
AK134589.1 | - | 12 | 1261 | ||
AK149654.1 | - | 11 | 1183 | ||
AK151438.1 | - | 11 | 1185 | ||
AK161613.1 | - | 11 | 1185 | ||
AK167588.1 | - | 11 | 1159 | ||
AK169199.1 | - | 11 | 1158 | ||
BC008997.1 | - | 11 | 1175 | 3157926 | |
L13129.1 | - | 11 | 1178 |