Gene Transcripts

Gene Symbol
Atp6v1e1
Entrez Gene ID
11973
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit E1
Gene Aliases
2410029D23Rik, Atp6e, Atp6e2, Atp6v1e, D6Ertd385e, E2, P31, Vma4
Chromosome Location
Chr. 6: 120795244 - 120822685 on Build GRCm38
UniGene ID
Mm.29045
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007510.2 NP_031536.2 2 111
GenBank mRNA AB074758.1 - 2 113
AK010603.1 - 2 128
AK139791.1 - 2 128
AK146162.1 - 2 125
AK149356.1 - 2 147
AK167644.1 - 2 70
AK169854.1 - 2 125
AK186366.1 - 2 63
AK189499.1 - 2 60
AK189578.1 - 2 63
BC003421.1 - 2 111 2598896
BC055438.1 - 2 145 3995744

View Full Product Details