Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015734.2 | NP_056549.2 | 4 | 895 | |
GenBank mRNA | AB009993.1 | - | 4 | 635 | |
BC138447.1 | - | 4 | 569 | 8861467 | |
BC138449.1 | - | 4 | 569 | 8861469 | |
S63661.1 | - | 3 | 399 |