Gene Transcripts

Gene Symbol
Diap1
Entrez Gene ID
13367
Gene Name
diaphanous homolog 1 (Drosophila)
Gene Aliases
D18Wsu154e, Dia1, Diaph1, Drf1, p140mDia
Chromosome Location
Chr. 18: 37843601 - 37935617 on Build GRCm38
UniGene ID
Mm.195916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK156391.1 - 11 1534
AK220466.1 - 24 3283
BC070412.1 - 25 3448 5705112
U96963.1 - 23 3286

View Full Product Details