Gene Transcripts

Gene Symbol
Epha6
Entrez Gene ID
13840
Gene Name
Eph receptor A6
Gene Aliases
Ehk2, Hek12, m-ehk2
Chromosome Location
Chr. 16: 59653061 - 60606484 on Build GRCm38
UniGene ID
Mm.455790
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007938.2 NP_031964.2 5 1659
GenBank mRNA BC141090.1 - 5 1175
BC145362.1 - 6 1295
U58332.1 - 5 1375

View Full Product Details