Gene Transcripts

Gene Symbol
Galc
Entrez Gene ID
14420
Gene Name
galactosylceramidase
Gene Aliases
2310068B06Rik, A930008M05Rik, AW212969, AW413532, Gacy, twi, twitcher
Chromosome Location
Chr. 12: 98202300 - 98259461 on Build GRCm38
UniGene ID
Mm.5120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK154760.1 - 14 1675
AK161339.1 - 9 965
BC065140.1 - 14 1547
BC086671.1 - 14 1552 30635559
D38557.1 - 14 1622

View Full Product Details