Gene Transcripts

Gene Symbol
Prl7a1
Entrez Gene ID
19113
Gene Name
prolactin family 7, subfamily a, member 1
Gene Aliases
AA407021, AA409971, AA410116, PLP-E, PLP-G, Prlpe, Prlpg
Chromosome Location
Chr. 13: 27633364 - 27642493 on Build GRCm38
UniGene ID
Mm.196424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164058.1 NP_001157530.1 6 524
NM_008930.2 NP_032956.1 6 521
GenBank mRNA AF011381.1 - 6 513
AF020525.1 - 6 495
AK028371.1 - 6 522
AK141309.1 - 6 525
AK146050.1 - 6 524
BB611867.1 - 6 522
BC051671.1 - 6 506 30018275

View Full Product Details