Gene Transcripts
- Gene Symbol
- Itgb4
- Entrez Gene ID
- 192897
- Gene Name
- integrin beta 4
- Gene Aliases
- AA407042, C230078O20, CD104
- Chromosome Location
- Chr. 11: 115974725 - 116008412 on Build GRCm38
- UniGene ID
- Mm.213873
- Species
- Mus musculus
- Gene Symbol
- B230344G16Rik
- Entrez Gene ID
- 100038636
- Gene Name
- RIKEN cDNA B230344G16 gene
- Gene Aliases
- ENSMUSG00000075418
- Chromosome Location
- Chr. 11: 115989722 - 115994110 on Build GRCm38
- UniGene ID
- Mm.450138
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK140207.1 |
- |
|
313 |
|
AK140292.1 |
- |
|
313 |
|
View Full Product Details