Gene Transcripts

Gene Symbol
Ranbp2
Entrez Gene ID
19386
Gene Name
RAN binding protein 2
Gene Aliases
A430087B05Rik, AI256741, NUP358
Chromosome Location
Chr. 10: 58446852 - 58494155 on Build GRCm38
UniGene ID
Mm.431695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011240.3 NP_035370.2 15 2200
GenBank mRNA AK145306.1 - 15 2131

View Full Product Details