Gene Transcripts

Gene Symbol
Rbpjl
Entrez Gene ID
19668
Gene Name
recombination signal binding protein for immunoglobulin kappa J region-like
Gene Aliases
RBP-L, Rbpsuhl
Chromosome Location
Chr. 2: 164403056 - 164415448 on Build GRCm38
UniGene ID
Mm.473794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009036.1 NP_033062.1 9 1004
GenBank mRNA AK052449.1 - 7 4115
AK131814.1 - 9 985
AK131832.1 - 9 941
BC152311.1 - 9 998 40090714
Y10926.1 - 9 1004

View Full Product Details