Gene Transcripts

Gene Symbol
Sf3a2
Entrez Gene ID
20222
Gene Name
splicing factor 3a, subunit 2
Gene Aliases
66kDa, AW047242, PRP11, SFA66, Sap62
Chromosome Location
Chr. 10: 80798664 - 80804924 on Build GRCm38
UniGene ID
Mm.358633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013651.4 NP_038679.3 6, 7 507
GenBank mRNA BC052697.1 - 6, 7 476
CX214888.1 - 1, 2 13

View Full Product Details